Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00895507_g1
          See other TSPOAP1 GE Assays ›
          Gene Symbol:
          TSPOAP1
          Gene Name:
          TSPO associated protein 1
          Gene Aliases:
          BZRAP1, PBR-IP, PRAX-1, PRAX1, RIM-BP1, RIMBP1
          Chromosome Location:
          Chr.17: 58301227 - 58329030 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00895507_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TSPOAP1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001261835.1 NP_001248764.1 26 - 27 6072 130
          NM_004758.3 NP_004749.2 26 - 27 6099 130
          NM_024418.2 NP_077729.1 25 - 26 5919 130
          XM_005257807.2 XP_005257864.1 26 - 27 6076 130
          XM_006722173.2 XP_006722236.1 26 - 27 6103 130
          XM_006722174.2 XP_006722237.1 25 - 26 6082 130
          XM_006722175.2 XP_006722238.1 25 - 26 5923 130
          XM_006722176.2 XP_006722239.1 25 - 26 5272 130
          XM_006722177.3 XP_006722240.1 22 - 23 4371 130
          XM_006722178.3 XP_006722241.1 21 - 22 4292 130
          XM_011525471.2 XP_011523773.1 14 - 15 3890 130
          XM_011525472.1 XP_011523774.1 16 - 17 4592 130
          XM_017025326.1 XP_016880815.1 26 - 27 6103 130
          XM_017025327.1 XP_016880816.1 27 - 28 5251 130
          XM_017025328.1 XP_016880817.1 24 - 25 5794 130
          XM_017025329.1 XP_016880818.1 25 - 26 5263 130
          XM_017025330.1 XP_016880819.1 25 - 26 5263 130
          XM_017025331.1 XP_016880820.1 25 - 26 5236 130
          XM_017025332.1 XP_016880821.1 24 - 25 5083 130
          XM_017025333.1 XP_016880822.1 24 - 25 4405 130
          XM_017025334.1 XP_016880823.1 23 - 24 4252 130
          GenBank mRNA AB014512.2 - 25 - 26 5888 130
          AF039571.1 - 26 - 27 5425 130
          BC146852.1 - 26 - 27 5392 9056780 130
          BC150279.1 - 25 - 26 5888 9007261 130
          BC171787.1 - 26 - 27 5365 9054441 130
          KJ534781.1 - 16 - 17 2303 130
          KJ534782.1 - 26 - 27 5201 130
          KJ534783.1 - 26 - 27 5228 130
          KJ534784.1 - 9 - 10 2408 130
          KJ534785.1 - 25 - 26 4361 130
          KJ534786.1 - 14 - 15 2534 130
          KJ534787.1 - 24 - 25 4208 130
          KJ534788.1 - 25 - 26 4388 130
          KJ534789.1 - 21 - 22 3113 130
          KJ534790.1 - 22 - 23 3269 130
          KJ535062.1 - 20 - 21 3163 130

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700011250.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17001728.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TSPOAP1

          Gene Ontology Categories:

          Function(s) Process(es)

          neurotransmitter secretion
          biological_process
          glutamate secretion
          protein binding
          benzodiazepine receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR17023154.hg.1 ENST00000268893.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17023154.hg.1 ENST00000343736.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17023154.hg.1 ENST00000355701.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17023154.hg.1 NM_004758.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17023154.hg.1 NM_024418.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17023154.hg.1 TC17001728.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1700190454.hg.1 ENST00000268893.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190454.hg.1 ENST00000343736.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190454.hg.1 ENST00000577871.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190454.hg.1 ENST00000578511.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190454.hg.1 ENST00000580669.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190454.hg.1 ENST00000581675.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190454.hg.1 NM_001261835.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700190454.hg.1 NM_004758.3 Refseq
          Affymetrix Human Clariom D Assay PSR1700190454.hg.1 NM_024418.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700190454.hg.1 TC1700011250.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700190454.hg.1 ENST00000268893.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190454.hg.1 ENST00000343736.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190454.hg.1 ENST00000577871.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190454.hg.1 ENST00000578511.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190454.hg.1 ENST00000580669.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190454.hg.1 ENST00000581675.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190454.hg.1 NM_001261835.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700190454.hg.1 NM_004758.3 Refseq
          Affymetrix Human Clariom D Assay PSR1700190454.hg.1 NM_024418.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0