Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00895508_g1
          See other TSPOAP1 GE Assays ›
          Gene Symbol:
          TSPOAP1
          Gene Name:
          TSPO associated protein 1
          Gene Aliases:
          BZRAP1, PBR-IP, PRAX-1, PRAX1, RIM-BP1, RIMBP1
          Chromosome Location:
          Chr.17: 58301227 - 58329030 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00895508_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TSPOAP1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001261835.1 NP_001248764.1 27 - 28 6100 111
          NM_004758.3 NP_004749.2 27 - 28 6127 111
          NM_024418.2 NP_077729.1 26 - 27 5947 111
          XM_005257807.2 XP_005257864.1 27 - 28 6104 111
          XM_006722173.2 XP_006722236.1 27 - 28 6131 111
          XM_006722174.2 XP_006722237.1 26 - 27 6110 111
          XM_006722175.2 XP_006722238.1 26 - 27 5951 111
          XM_006722176.2 XP_006722239.1 26 - 27 5300 111
          XM_006722177.3 XP_006722240.1 23 - 24 4399 111
          XM_006722178.3 XP_006722241.1 22 - 23 4320 111
          XM_011525471.2 XP_011523773.1 15 - 16 3918 111
          XM_011525472.1 XP_011523774.1 17 - 18 4620 111
          XM_017025326.1 XP_016880815.1 27 - 28 6131 111
          XM_017025327.1 XP_016880816.1 28 - 29 5279 111
          XM_017025328.1 XP_016880817.1 25 - 26 5822 111
          XM_017025329.1 XP_016880818.1 26 - 27 5291 111
          XM_017025330.1 XP_016880819.1 26 - 27 5291 111
          XM_017025331.1 XP_016880820.1 26 - 27 5264 111
          XM_017025332.1 XP_016880821.1 25 - 26 5111 111
          XM_017025333.1 XP_016880822.1 25 - 26 4433 111
          XM_017025334.1 XP_016880823.1 24 - 25 4280 111
          GenBank mRNA AB014512.2 - 26 - 27 5916 111
          AF039571.1 - 27 - 28 5453 111
          BC031401.1 - 2 - 3 48 111
          BC146852.1 - 27 - 28 5420 9056780 111
          BC150279.1 - 26 - 27 5916 9007261 111
          BC171787.1 - 27 - 28 5393 9054441 111
          KJ534781.1 - 17 - 18 2331 111
          KJ534782.1 - 27 - 28 5229 111
          KJ534783.1 - 27 - 28 5256 111
          KJ534784.1 - 10 - 11 2436 111
          KJ534785.1 - 26 - 27 4389 111
          KJ534786.1 - 15 - 16 2562 111
          KJ534787.1 - 25 - 26 4236 111
          KJ534788.1 - 26 - 27 4416 111
          KJ534789.1 - 22 - 23 3141 111
          KJ534790.1 - 23 - 24 3297 111
          KJ535062.1 - 21 - 22 3191 111

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700011250.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17001728.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TSPOAP1

          Gene Ontology Categories:

          Function(s) Process(es)

          neurotransmitter secretion
          biological_process
          glutamate secretion
          protein binding
          benzodiazepine receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR17023153.hg.1 ENST00000268893.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17023153.hg.1 ENST00000343736.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17023153.hg.1 ENST00000355701.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17023153.hg.1 NM_004758.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17023153.hg.1 NM_024418.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17023153.hg.1 TC17001728.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1700190453.hg.1 ENST00000268893.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190453.hg.1 ENST00000343736.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190453.hg.1 ENST00000577871.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190453.hg.1 ENST00000578511.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190453.hg.1 ENST00000580669.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190453.hg.1 ENST00000581675.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190453.hg.1 NM_001261835.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700190453.hg.1 NM_004758.3 Refseq
          Affymetrix Human Clariom D Assay PSR1700190453.hg.1 NM_024418.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700190453.hg.1 TC1700011250.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700190453.hg.1 ENST00000268893.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190453.hg.1 ENST00000343736.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190453.hg.1 ENST00000577871.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190453.hg.1 ENST00000578511.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190453.hg.1 ENST00000580669.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190453.hg.1 ENST00000581675.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190453.hg.1 NM_001261835.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700190453.hg.1 NM_004758.3 Refseq
          Affymetrix Human Clariom D Assay PSR1700190453.hg.1 NM_024418.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0