Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00900061_m1
          See other HGF GE Assays ›
          Gene Symbol:
          HGF
          Gene Name:
          hepatocyte growth factor
          Gene Aliases:
          DFNB39, F-TCF, HGFB, HPTA, SF
          Chromosome Location:
          Chr.7: 81699006 - 81770438 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00900061_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HGF
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000601.5 NP_000592.3 12 - 13 1666 108
          NM_001010932.2 NP_001010932.1 12 - 13 1651 108
          XM_006715956.2 XP_006716019.1 13 - 14 1629 108
          XM_011516115.2 XP_011514417.1 13 - 14 1614 108
          GenBank mRNA AK291674.1 - 12 - 13 1601 108
          AK313694.1 - 12 - 13 1501 108
          BC130284.1 - 12 - 13 1514 40146315 108
          BC130286.1 - 12 - 13 1514 40146317 108
          M29145.1 - 12 - 13 1540 108
          M55379.1 - 12 - 13 1424 108
          M60718.1 - 12 - 13 1511 108
          M73239.1 - 12 - 13 1504 108
          M73240.1 - 12 - 13 1489 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700011642.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07001562.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HGF

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          epithelial to mesenchymal transition
          liver development
          platelet degranulation
          proteolysis
          positive regulation of cell population proliferation
          negative regulation of autophagy
          positive regulation of phosphatidylinositol 3-kinase signaling
          hyaluronan metabolic process
          positive regulation of cell migration
          animal organ regeneration
          positive regulation of myelination
          negative regulation of interleukin-6 production
          positive regulation of interleukin-10 production
          negative regulation of peptidyl-serine phosphorylation
          cellular response to hepatocyte growth factor stimulus
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of osteoblast differentiation
          positive regulation of angiogenesis
          positive regulation of transcription by RNA polymerase II
          hepatocyte growth factor receptor signaling pathway
          negative regulation of inflammatory response
          positive regulation of peptidyl-tyrosine phosphorylation
          positive chemotaxis
          myoblast proliferation
          cell chemotaxis
          regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling
          positive regulation of neuron projection regeneration
          negative regulation of release of cytochrome c from mitochondria
          regulation of p38MAPK cascade
          negative regulation of hydrogen peroxide-mediated programmed cell death
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          regulation of tau-protein kinase activity
          positive regulation of DNA biosynthetic process
          serine-type endopeptidase activity
          protein binding
          growth factor activity
          chemoattractant activity
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR07023475.hg.1 ENST00000222390.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07023475.hg.1 ENST00000457544.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07023475.hg.1 NM_000601.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07023475.hg.1 NM_001010932.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07023475.hg.1 TC07001562.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0700183021.hg.1 ENST00000222390.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0700183021.hg.1 ENST00000457544.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0700183021.hg.1 NM_000601.5 Refseq
          Affymetrix Human Clariom D Assay PSR0700183021.hg.1 NM_001010932.2 Refseq
          Affymetrix Human Clariom D Assay PSR0700183021.hg.1 TC0700011642.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700183021.hg.1 ENST00000222390.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0700183021.hg.1 ENST00000457544.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0700183021.hg.1 NM_000601.5 Refseq
          Affymetrix Human Clariom D Assay PSR0700183021.hg.1 NM_001010932.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0