Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00900503_m1
          See other PELI1 GE Assays ›
          Gene Symbol:
          PELI1
          Gene Name:
          pellino E3 ubiquitin protein ligase 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 64092652 - 64144471 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00900503_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PELI1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020651.3 NP_065702.2 4 - 5 766 110
          XM_005264437.3 XP_005264494.1 3 - 4 585 110
          XM_011532994.2 XP_011531296.1 4 - 5 1425 110
          XM_011532996.2 XP_011531298.1 4 - 5 2184 110
          XM_017004520.1 XP_016860009.1 5 - 6 900 110
          XM_017004521.1 XP_016860010.1 5 - 6 759 110
          GenBank mRNA AF300987.1 - 3 - 4 321 110
          AF302505.1 - 3 - 4 4344 110
          AJ278859.1 - 3 - 4 306 110
          AK312441.1 - 4 - 5 715 110
          AY364257.1 - 3 - 4 306 110
          BC050019.1 - 4 - 5 720 5296990 110
          BC050533.1 - 4 - 5 702 110
          BC063611.1 - 4 - 5 694 4992160 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200012809.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02001911.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the XM_005264437.3, XM_011532996.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PELI1

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cytokine production
          Toll signaling pathway
          positive regulation of B cell proliferation
          positive regulation of protein ubiquitination
          negative regulation of NF-kappaB transcription factor activity
          response to lipopolysaccharide
          positive regulation of toll-like receptor 3 signaling pathway
          positive regulation of toll-like receptor 4 signaling pathway
          negative regulation of T cell proliferation
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          response to dsRNA
          protein K48-linked ubiquitination
          protein binding
          ligase activity
          ubiquitin-ubiquitin ligase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000468869.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000468869.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000468869.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000468869.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02030568.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02030568.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02030568.hg.1 TC02001911.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02030567.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02030567.hg.1 NM_020651.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02030567.hg.1 TC02001911.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000466177.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000468869.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000468869.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000468869.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000468869.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 ENST00000494203.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012809.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200193433.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193433.hg.1 NM_020651.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200193433.hg.1 TC0200012809.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200193433.hg.1 ENST00000358912.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193433.hg.1 NM_020651.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7f58f9545f-46b6w:80/100.66.130.62:80.
          git-commit: c36c52929dfaa9f2956040e6abd275853d6e1b95
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline