Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00902207_g1
          See other SYCP2 GE Assays ›
          Gene Symbol:
          SYCP2
          Gene Name:
          synaptonemal complex protein 2
          Gene Aliases:
          SCP-2, SCP2
          Chromosome Location:
          Chr.20: 59863277 - 59933700 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00902207_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SYCP2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_014258.3 NP_055073.2 22 - 23 2021 96
          XM_011528487.2 XP_011526789.1 22 - 23 2057 96
          XM_011528488.2 XP_011526790.1 22 - 23 2009 96
          XM_011528489.2 XP_011526791.1 22 - 23 2052 96
          XM_011528490.2 XP_011526792.1 22 - 23 2033 96
          XM_011528493.2 XP_011526795.1 21 - 22 1972 96
          XM_017027586.1 XP_016883075.1 21 - 22 2253 96
          XM_017027587.1 XP_016883076.1 21 - 22 2250 96
          XM_017027588.1 XP_016883077.1 21 - 22 2254 96
          XM_017027589.1 XP_016883078.1 21 - 22 2254 96
          XM_017027590.1 XP_016883079.1 21 - 22 1946 96
          XM_017027591.1 XP_016883080.1 13 - 14 1352 96
          XM_017027592.1 XP_016883081.1 21 - 22 2255 96
          GenBank mRNA AL080226.1 - 10 - 11 955 96
          BC040566.1 - 22 - 23 2057 96
          BC132864.1 - 21 - 22 1890 40146886 96
          BC132870.1 - 21 - 22 1892 40146892 96
          BX648091.1 - 22 - 23 1975 96
          BX649108.1 - 22 - 23 2001 96
          Y08982.1 - 20 - 21 1800 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000009673.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000999.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SYCP2

          Gene Ontology Categories:

          Function(s) Process(es)

          synaptonemal complex assembly
          male meiotic nuclear division
          female meiotic nuclear division
          fertilization
          negative regulation of apoptotic process
          male genitalia morphogenesis
          cell division
          DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR20013605.hg.1 TC20000999.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20013605.hg.1 ENST00000357552.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20013605.hg.1 ENST00000371001.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20013605.hg.1 NM_014258.3 Refseq
          Affymetrix Human Clariom D Assay PSR2000167491.hg.1 ENST00000357552.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2000167491.hg.1 ENST00000371001.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2000167491.hg.1 ENST00000446834.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2000167491.hg.1 NM_014258.3 Refseq
          Affymetrix Human Clariom D Assay PSR2000167491.hg.1 TC2000009673.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000167491.hg.1 NM_014258.3 Refseq
          Affymetrix Human Clariom D Assay PSR2000167491.hg.1 ENST00000357552.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2000167491.hg.1 ENST00000371001.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2000167491.hg.1 ENST00000446834.5 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0