Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021405.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021405.hg.1 |
ENST00000443356.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021405.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021405.hg.1 |
TC08002620.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012467.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188162.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188162.hg.1 |
NM_001080399.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188162.hg.1 |
TC0800012467.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800188162.hg.1 |
ENST00000254627.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188162.hg.1 |
NM_001080399.2 |
Refseq |