Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00903489_g1
          See other ANP32B GE Assays ›
          Gene Symbol:
          ANP32B
          Gene Name:
          acidic nuclear phosphoprotein 32 family member B
          Gene Aliases:
          APRIL, PHAPI2, SSP29
          Chromosome Location:
          Chr.9: 97983207 - 98015943 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00903489_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ANP32B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_006401.2 NP_006392.1 1 - 2 402 105
          GenBank mRNA AK223454.1 - 1 - 2 269 105
          AK313733.1 - 1 - 2 257 105
          BC013003.2 - 1 - 2 234 3010189 105
          BC019658.1 - 1 - 2 241 4478860 105
          U70439.1 - 1 - 2 263 105
          Y07569.1 - 1 - 2 108 105
          Y07570.1 - 1 - 2 237 105
          Y07969.1 - 1 - 2 277 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900008173.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09000495.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ANP32B

          Gene Ontology Categories:

          Function(s) Process(es)

          vasculature development
          nucleosome assembly
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          ventricular system development
          negative regulation of cell differentiation
          positive regulation of protein export from nucleus
          inner ear development
          roof of mouth development
          protein binding
          histone binding
          RNA polymerase binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 ENST00000486769.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 ENST00000486769.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 ENST00000486769.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09005349.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09005349.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09005349.hg.1 NM_006401.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09005349.hg.1 TC09000495.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09005354.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09005354.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09005354.hg.1 NM_006401.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09005354.hg.1 TC09000495.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1500010212.hg.1 ENST00000559213.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09005342.hg.1 NM_006401.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09005342.hg.1 TC09000495.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09005351.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09005351.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09005351.hg.1 NM_006401.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09005351.hg.1 TC09000495.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09005346.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09005346.hg.1 NM_006401.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09005346.hg.1 TC09000495.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09005350.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09005350.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09005350.hg.1 NM_006401.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09005350.hg.1 TC09000495.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2200007594.hg.1 ENST00000454525.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09005347.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09005347.hg.1 NM_006401.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09005347.hg.1 TC09000495.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1500010068.hg.1 ENST00000564205.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010068.hg.1 ENST00000564205.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010068.hg.1 ENST00000564205.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010068.hg.1 ENST00000568553.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154784.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154784.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154784.hg.1 ENST00000486769.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154784.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900154784.hg.1 TC0900008173.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900154784.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900154784.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154784.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154784.hg.1 ENST00000486769.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500010212.hg.1 ENST00000559213.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154773.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154773.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900154773.hg.1 TC0900008173.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900154773.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900154773.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154769.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900154769.hg.1 TC0900008173.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900154769.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 ENST00000486769.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 ENST00000486769.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 ENST00000486769.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008173.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007594.hg.1 ENST00000454525.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154772.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154772.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900154772.hg.1 TC0900008173.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900154772.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154772.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900154778.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154778.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154778.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900154778.hg.1 TC0900008173.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900154778.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154778.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154778.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900154776.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154776.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154776.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900154776.hg.1 TC0900008173.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900154776.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154776.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154776.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay TC1500010068.hg.1 ENST00000564205.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500010068.hg.1 ENST00000564205.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500010068.hg.1 ENST00000564205.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500010068.hg.1 ENST00000568553.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154779.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154779.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154779.hg.1 ENST00000486769.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154779.hg.1 NM_006401.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900154779.hg.1 TC0900008173.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900154779.hg.1 ENST00000339399.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154779.hg.1 ENST00000473205.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154779.hg.1 ENST00000486769.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154779.hg.1 NM_006401.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 173406 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0