Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00906878_m1
          See other NDRG1 GE Assays ›
          Gene Symbol:
          NDRG1
          Gene Name:
          N-myc downstream regulated 1
          Gene Aliases:
          CAP43, CMT4D, DRG-1, DRG1, GC4, HMSNL, NDR1, NMSL, PROXY1, RIT42, RTP, TARG1, TDD5
          Chromosome Location:
          Chr.8: 133237171 - 133297587 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00906878_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NDRG1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135242.1 NP_001128714.1 7 - 8 1038 62
          NM_001258432.1 NP_001245361.1 5 - 6 524 62
          NM_001258433.1 NP_001245362.1 6 - 7 535 62
          NM_006096.3 NP_006087.2 7 - 8 641 62
          XM_011516791.1 XP_011515093.1 7 - 8 924 62
          GenBank mRNA AF004162.1 - 6 - 7 480 62
          AK091147.1 - 6 - 7 484 62
          AK096562.1 - 7 - 8 476 62
          AK126924.1 - 7 - 8 1320 62
          AK223542.1 - 7 - 8 543 62
          AK296794.1 - 5 - 6 472 62
          AK297342.1 - 772 62
          AK307829.1 - 7 - 8 590 62
          AU141826.1 - 7 - 8 593 62
          AY277597.1 - 6 - 7 469 62
          BC003175.1 - 7 - 8 569 2957847 62
          CR456842.1 - 6 - 7 452 62
          D87953.1 - 7 - 8 574 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as HTA transcript cluster(s) TC08001659.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NDRG1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of cell population proliferation
          response to metal ion
          DNA damage response, signal transduction by p53 class mediator
          peripheral nervous system myelin maintenance
          regulation of apoptotic process
          mast cell activation
          cellular response to hypoxia
          positive regulation of spindle checkpoint
          cell-cell adhesion
          protein binding
          microtubule binding
          gamma-tubulin binding
          cadherin binding
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR08021642.hg.1 ENST00000323851.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021642.hg.1 ENST00000354944.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021642.hg.1 ENST00000414097.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021642.hg.1 ENST00000522476.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021642.hg.1 ENST00000537882.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021642.hg.1 NM_001135242.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08021642.hg.1 NM_006096.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08021642.hg.1 TC08001659.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08021643.hg.1 ENST00000323851.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021643.hg.1 ENST00000354944.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021643.hg.1 ENST00000414097.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021643.hg.1 ENST00000522476.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021643.hg.1 ENST00000535532.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021643.hg.1 ENST00000537882.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021643.hg.1 NM_001135242.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08021643.hg.1 NM_006096.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08021643.hg.1 TC08001659.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08021644.hg.1 ENST00000323851.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021644.hg.1 ENST00000354944.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021644.hg.1 ENST00000414097.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021644.hg.1 ENST00000522476.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021644.hg.1 ENST00000537882.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021644.hg.1 NM_001135242.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08021644.hg.1 NM_006096.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08021644.hg.1 TC08001659.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08021645.hg.1 ENST00000323851.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021645.hg.1 ENST00000354944.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021645.hg.1 ENST00000414097.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021645.hg.1 ENST00000522476.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021645.hg.1 ENST00000537882.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021645.hg.1 NM_001135242.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08021645.hg.1 NM_006096.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08021645.hg.1 TC08001659.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08021648.hg.1 ENST00000323851.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021648.hg.1 ENST00000354944.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021648.hg.1 ENST00000414097.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021648.hg.1 ENST00000522476.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021648.hg.1 ENST00000537882.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021648.hg.1 NM_001135242.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08021648.hg.1 NM_006096.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08021648.hg.1 TC08001659.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08021653.hg.1 ENST00000323851.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021653.hg.1 ENST00000354944.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021653.hg.1 ENST00000414097.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021653.hg.1 ENST00000522476.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021653.hg.1 ENST00000537882.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021653.hg.1 NM_001135242.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08021653.hg.1 NM_006096.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08021653.hg.1 TC08001659.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08021646.hg.1 ENST00000323851.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021646.hg.1 ENST00000354944.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021646.hg.1 ENST00000414097.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021646.hg.1 ENST00000522476.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021646.hg.1 ENST00000537882.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021646.hg.1 NM_001135242.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08021646.hg.1 NM_006096.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08021646.hg.1 TC08001659.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08021647.hg.1 ENST00000323851.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021647.hg.1 ENST00000354944.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021647.hg.1 ENST00000414097.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021647.hg.1 ENST00000522476.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021647.hg.1 ENST00000537882.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021647.hg.1 NM_001135242.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08021647.hg.1 NM_006096.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08021647.hg.1 TC08001659.hg.1 HTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0