Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00907322_m1
          See other AHR GE Assays ›
          Gene Symbol:
          AHR
          Gene Name:
          aryl hydrocarbon receptor
          Gene Aliases:
          bHLHe76
          Chromosome Location:
          Chr.7: 17298652 - 17346151 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00907322_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AHR
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001621.4 NP_001612.1 9 - 10 1768 146
          GenBank mRNA AK307495.1 - 9 - 10 1768 146
          BC069390.1 - 9 - 10 1225 7262381 146
          BC070080.1 - 9 - 10 1686 30342582 146
          D16354.1 - 9 - 10 1737 146
          L19872.1 - 9 - 10 1530 146

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700006795.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07000112.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene AHR

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle development
          blood vessel development
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          xenobiotic metabolic process
          apoptotic process
          cell cycle
          response to xenobiotic stimulus
          response to toxic substance
          regulation of gene expression
          intracellular receptor signaling pathway
          regulation of B cell proliferation
          response to estradiol
          circadian regulation of gene expression
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          cellular response to glucocorticoid stimulus
          transcription cis-regulatory region binding
          cis-regulatory region sequence-specific DNA binding
          DNA binding
          DNA-binding transcription factor activity
          nuclear receptor activity
          protein binding
          transcription factor binding
          protein heterodimerization activity
          protein dimerization activity
          Hsp90 protein binding
          E-box binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR07001739.hg.1 ENST00000242057.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07001739.hg.1 ENST00000492120.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07001739.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07001739.hg.1 TC07000112.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000475440.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000475440.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000475440.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000475440.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000481944.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000481944.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000481944.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000492120.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000492120.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07001740.hg.1 ENST00000242057.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07001740.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07001740.hg.1 TC07000112.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0700146971.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0700146971.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700146971.hg.1 ENST00000492120.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700146971.hg.1 NM_001621.4 Refseq
          Affymetrix Human Clariom D Assay PSR0700146971.hg.1 TC0700006795.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700146971.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0700146971.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700146971.hg.1 ENST00000492120.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700146971.hg.1 NM_001621.4 Refseq
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000475440.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000475440.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000475440.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000475440.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000481944.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000481944.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000481944.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000492120.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 ENST00000492120.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Clariom D Assay TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Clariom D Assay PSR0700146972.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0700146972.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700146972.hg.1 NM_001621.4 Refseq
          Affymetrix Human Clariom D Assay PSR0700146972.hg.1 TC0700006795.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700146972.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0700146972.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700146972.hg.1 NM_001621.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline