Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00907951_m1
          See other PIK3CA GE Assays ›
          Gene Symbol:
          PIK3CA
          Gene Name:
          phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha
          Gene Aliases:
          CLOVE, CWS5, MCAP, MCM, MCMTC, PI3K, PI3K-alpha, p110-alpha
          Chromosome Location:
          Chr.3: 179148114 - 179240093 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00907951_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PIK3CA
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_006218.3 NP_006209.2 14 - 15 2350 109
          XM_006713658.3 XP_006713721.1 14 - 15 2375 109
          XM_011512894.2 XP_011511196.1 14 - 15 2456 109
          GenBank mRNA AB210020.1 - 13 - 14 2198 109
          AK292940.1 - 14 - 15 2308 109
          AK297203.1 - 11 - 12 1589 109
          AK302608.1 - 13 - 14 2028 109
          BC113601.1 - 13 - 14 2270 8322653 109
          BC113603.1 - 13 - 14 2270 8322655 109
          U79143.1 - 13 - 14 2193 109
          Z29090.1 - 13 - 14 2205 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300009597.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03000951.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PIK3CA

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          liver development
          vasculature development
          glucose metabolic process
          protein phosphorylation
          phosphatidylinositol biosynthetic process
          epidermal growth factor receptor signaling pathway
          regulation of gene expression
          regulation of phosphatidylinositol 3-kinase signaling
          platelet activation
          T cell costimulation
          activation of protein kinase activity
          positive regulation of peptidyl-serine phosphorylation
          phosphatidylinositol-3-phosphate biosynthetic process
          insulin receptor signaling pathway via phosphatidylinositol 3-kinase
          Fc-epsilon receptor signaling pathway
          Fc-gamma receptor signaling pathway involved in phagocytosis
          ERBB2 signaling pathway
          regulation of multicellular organism growth
          regulation of cellular respiration
          protein kinase B signaling
          negative regulation of neuron apoptotic process
          endothelial cell migration
          hypomethylation of CpG island
          phosphatidylinositol phosphate biosynthetic process
          vascular endothelial growth factor receptor signaling pathway
          phosphatidylinositol-mediated signaling
          T cell receptor signaling pathway
          leukocyte migration
          cardiac muscle contraction
          adipose tissue development
          cellular response to glucose stimulus
          energy homeostasis
          positive regulation of mitophagy in response to mitochondrial depolarization
          negative regulation of fibroblast apoptotic process
          regulation of genetic imprinting
          negative regulation of anoikis
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          kinase activity
          1-phosphatidylinositol-3-kinase activity
          protein kinase activator activity
          phosphatidylinositol 3-kinase activity
          1-phosphatidylinositol-4-phosphate 3-kinase activity
          insulin receptor substrate binding
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2200006480.hg.1 ENST00000454360.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006480.hg.1 ENST00000454360.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006480.hg.1 ENST00000454360.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006480.hg.1 ENST00000454360.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006480.hg.1 ENST00000454360.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006480.hg.1 ENST00000454360.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000462255.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000462255.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000462255.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000462255.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000462255.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000462255.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000468036.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000468036.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000468036.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000468036.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000477735.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000477735.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 ENST00000477735.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03016381.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03016381.hg.1 NM_006218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03016381.hg.1 TC03000951.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC2200006480.hg.1 ENST00000454360.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200006480.hg.1 ENST00000454360.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200006480.hg.1 ENST00000454360.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200006480.hg.1 ENST00000454360.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200006480.hg.1 ENST00000454360.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200006480.hg.1 ENST00000454360.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300168736.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300168736.hg.1 ENST00000462255.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300168736.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300168736.hg.1 TC0300009597.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300168736.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300168736.hg.1 ENST00000462255.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300168736.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000263967.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000462255.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000462255.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000462255.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000462255.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000462255.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000462255.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000468036.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000468036.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000468036.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000468036.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000477735.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000477735.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 ENST00000477735.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq
          Affymetrix Human Clariom D Assay TC0300009597.hg.1 NM_006218.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0