Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00908810_g1
          See other KAT2B GE Assays ›
          Gene Symbol:
          KAT2B
          Gene Name:
          lysine acetyltransferase 2B
          Gene Aliases:
          CAF, P/CAF, PCAF
          Chromosome Location:
          Chr.3: 20040032 - 20154404 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00908810_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KAT2B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003884.4 NP_003875.3 16 - 17 2664 56
          XM_017007423.1 XP_016862912.1 17 - 18 2119 56
          XM_017007424.1 XP_016862913.1 16 - 17 2215 56
          GenBank mRNA AL832173.1 - 6 - 7 577 56
          BC060823.1 - 16 - 17 2269 30336561 56
          BC070075.1 - 16 - 17 2334 30333414 56
          U57317.2 - 16 - 17 2676 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300006791.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03000113.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene KAT2B

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin remodeling
          transcription initiation from RNA polymerase II promoter
          protein acetylation
          Notch signaling pathway
          memory
          negative regulation of cell population proliferation
          regulation of protein ADP-ribosylation
          positive regulation of neuron projection development
          viral process
          N-terminal peptidyl-lysine acetylation
          internal peptidyl-lysine acetylation
          peptidyl-lysine acetylation
          cellular response to insulin stimulus
          cellular response to oxidative stress
          positive regulation of chromatin binding
          histone H3 acetylation
          histone H3-K9 acetylation
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          positive regulation of gene expression, epigenetic
          positive regulation of transcription by RNA polymerase II
          rhythmic process
          cellular response to parathyroid hormone stimulus
          positive regulation of histone H3-K14 acetylation
          positive regulation of histone H3-K9 acetylation
          RNA polymerase II transcription regulatory region sequence-specific DNA binding
          chromatin binding
          transcription coregulator activity
          transcription coactivator activity
          histone acetyltransferase activity
          lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
          cyclin-dependent protein serine/threonine kinase inhibitor activity
          protein binding
          transcription factor binding
          acetyltransferase activity
          protein kinase binding
          histone acetyltransferase binding
          histone deacetylase binding
          protein-containing complex binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR03002305.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03002305.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03002305.hg.1 TC03000113.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000426228.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000426228.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000426228.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000426228.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000426228.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000468111.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000468111.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000468111.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000468111.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000468111.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000468400.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000468400.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000468400.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000469085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000469085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000469085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 ENST00000469085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000426228.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000426228.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000426228.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000426228.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000426228.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000468111.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000468111.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000468111.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000468111.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000468111.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000468400.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000468400.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000468400.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000469085.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000469085.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000469085.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 ENST00000469085.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay TC0300006791.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay PSR0300147474.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300147474.hg.1 ENST00000468400.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300147474.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay PSR0300147474.hg.1 TC0300006791.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300147474.hg.1 NM_003884.4 Refseq
          Affymetrix Human Clariom D Assay PSR0300147474.hg.1 ENST00000263754.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300147474.hg.1 ENST00000468400.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-tbs5g:80/100.66.134.240:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0