Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000483182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000483182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000483182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000483182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000483182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000483182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000483182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000483182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000486563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000486563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000486563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000486563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000486563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000486563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000488453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000488453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000488453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000488453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000488453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000488453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000488453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000488453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000488453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
ENST00000488453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007467.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007467.hg.1 |
ENST00000488453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007467.hg.1 |
ENST00000537711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007467.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007467.hg.1 |
TC01000484.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0100155323.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155323.hg.1 |
ENST00000483182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155323.hg.1 |
ENST00000488453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155323.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155323.hg.1 |
TC0100007858.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100155323.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155323.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155323.hg.1 |
ENST00000483182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155323.hg.1 |
ENST00000488453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000373014.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000483182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000483182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000483182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000483182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000483182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000483182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000483182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000483182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000486563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000486563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000486563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000486563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000486563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000486563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000488453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000488453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000488453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000488453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000488453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000488453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000488453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000488453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000488453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
ENST00000488453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007858.hg.1 |
NM_016037.3 |
Refseq |