Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00917374_s1
          See other URB1-AS1 GE Assays ›
          Gene Symbol:
          URB1-AS1
          Gene Name:
          URB1 antisense RNA 1 (head to head)
          Gene Aliases:
          C21orf119, PRED84
          Chromosome Location:
          Chr.21: 32393133 - 32393960 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00917374_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: URB1-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026845.1 1 744 126
          GenBank mRNA AI369236.1 - 1 126
          AK026376.1 - 1 722 126
          BC007928.1 - 1 750 126
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AW024805.1 - 1 Chr. 21 - 32393489 - 32393951 [-] on Build GRCh38 126
          GenBankmRNA DB567217.1 - 1 382 Chr. 21 - 32393495 - 32393951 [+] on Build GRCh38 126
          GenBankmRNA AI004809.1 - 1 Chr. 21 - 32393586 - 32393960 [-] on Build GRCh38 126
          GenBankmRNA BE673100.1 - 1 Chr. 21 - 32393496 - 32393958 [-] on Build GRCh38 126
          GenBankmRNA AI769042.1 - 1 Chr. 21 - 32393516 - 32393950 [-] on Build GRCh38 126
          GenBankmRNA AI290359.1 - 1 Chr. 21 - 32393487 - 32393955 [-] on Build GRCh38 126
          GenBankmRNA AI873229.1 - 1 Chr. 21 - 32393665 - 32393951 [-] on Build GRCh38 126
          GenBankmRNA AI572165.1 - 1 Chr. 21 - 32393666 - 32393956 [-] on Build GRCh38 126
          GenBankmRNA AI800837.1 - 1 Chr. 21 - 32393398 - 32393960 [-] on Build GRCh38 126
          GenBankmRNA AI268567.1 - 1 Chr. 21 - 32393546 - 32393953 [-] on Build GRCh38 126
          GenBankmRNA AI335938.1 - 1 Chr. 21 - 32393520 - 32393941 [-] on Build GRCh38 126
          GenBankmRNA BI756790.1 - 1 721 Chr. 21 - 32393158 - 32393962 [+] on Build GRCh38 126
          GenBankmRNA AI265972.1 - 1 Chr. 21 - 32393618 - 32393960 [-] on Build GRCh38 126
          GenBankmRNA AW973865.1 - 1 Chr. 21 - 32393359 - 32393944 [-] on Build GRCh38 126
          GenBankmRNA AI336791.1 - 1 Chr. 21 - 32393546 - 32393958 [-] on Build GRCh38 126
          GenBankmRNA AA831850.1 - 1 Chr. 21 - 32393619 - 32393962 [-] on Build GRCh38 126
          GenBankmRNA AI299419.1 - 1 Chr. 21 - 32393530 - 32393962 [-] on Build GRCh38 126
          GenBankmRNA BU728745.1 - 1 Chr. 21 - 32393501 - 32393962 [-] on Build GRCh38 126
          GenBankmRNA AW663472.1 - 1 Chr. 21 - 32393530 - 32393959 [-] on Build GRCh38 126
          GenBankmRNA AI276483.1 - 1 Chr. 21 - 32393529 - 32393959 [-] on Build GRCh38 126
          GenBankmRNA AI887961.1 - 1 Chr. 21 - 32393483 - 32393955 [-] on Build GRCh38 126
          GenBankmRNA AI291328.1 - 1 Chr. 21 - 32393521 - 32393962 [-] on Build GRCh38 126
          GenBankmRNA BE504401.1 - 1 Chr. 21 - 32393489 - 32393955 [-] on Build GRCh38 126
          GenBankmRNA BG055225.1 - 1 Chr. 21 - 32393624 - 32393951 [-] on Build GRCh38 126
          GenBankmRNA BM987277.1 - 1 Chr. 21 - 32393501 - 32393962 [-] on Build GRCh38 126
          GenBankmRNA W72429.1 - Chr. - - [-] on Build 126
          GenBankmRNA AI338974.1 - 1 Chr. 21 - 32393495 - 32393956 [-] on Build GRCh38 126
          GenBankmRNA AI377725.1 - 1 Chr. 21 - 32393545 - 32393958 [-] on Build GRCh38 126
          GenBankmRNA AI335928.1 - 1 Chr. 21 - 32393539 - 32393941 [-] on Build GRCh38 126
          GenBankmRNA AI362878.1 - 1 Chr. 21 - 32393545 - 32393958 [-] on Build GRCh38 126
          GenBankmRNA BM692195.1 - 1 400 Chr. 21 - 32393485 - 32393944 [+] on Build GRCh38 126

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream URB1 ribosome biogenesis 1 homolog (S. cerevisiae) URB1 C21orf108,
          NPA1
          -513.0 Sense Chr. 21 - 32311019 - 32393003 on NCBI Build GRCh38
          Downstream eva-1 homolog C (C. elegans) EVA1C B18,
          B19,
          C21orf63,
          C21orf64,
          FAM176C,
          PRED34,
          SUE21
          17.7kb Anti-sense Chr. 21 - 32411692 - 32515400 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Process(es)

          protein binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11354580 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32014194 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32014190 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32014187 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32014192 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17231178 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17231177 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32014196 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11354576 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11354575 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11354589 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11354588 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11354583 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11354587 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 27328 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 48028 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P29029 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 12389 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 13204 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 4458 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P29029 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P29029 - -
          Agilent Whole Chr array_18 19 21 A_17_P17231177 - -
          Agilent Whole Chr array_18 19 21 A_17_P32014190 - -
          Agilent Whole Chr array_18 19 21 A_17_P11354575 - -
          Agilent Whole Chr array_18 19 21 A_17_P11354589 - -
          Agilent Whole Chr array_18 19 21 A_17_P11354577 - -
          Agilent Whole Chr array_18 19 21 A_17_P17231176 - -
          Agilent Whole Chr array_18 19 21 A_17_P11354588 - -
          Agilent Whole Chr array_18 19 21 A_17_P32014205 - -
          Agilent Whole Chr array_18 19 21 A_17_P17231175 - -
          Agilent Whole Chr array_18 19 21 A_17_P11354584 - -
          Agilent Whole Chr array_18 19 21 A_17_P32014194 - -
          Agilent Whole Chr array_18 19 21 A_17_P17231178 - -
          Agilent Whole Chr array_18 19 21 A_17_P32014193 - -
          Agilent Whole Chr array_18 19 21 A_17_P32014201 - -
          Agilent Whole Chr array_18 19 21 A_17_P11354587 - -
          Agilent Whole Chr array_18 19 21 A_17_P11354576 - -
          Agilent Whole Chr array_18 19 21 A_17_P32014203 - -
          Agilent Whole Chr array_18 19 21 A_17_P11354590 - -
          Agilent Whole Chr array_18 19 21 A_17_P32014192 - -
          Agilent Whole Chr array_18 19 21 A_17_P32014187 - -
          Agilent Whole Chr array_18 19 21 A_17_P32014197 - -
          Agilent Whole Chr array_18 19 21 A_17_P11354581 - -
          Agilent Whole Chr array_18 19 21 A_17_P32014196 - -
          Agilent Whole Chr array_18 19 21 A_17_P11354580 - -
          Agilent Whole Chr array_18 19 21 A_17_P11354583 - -
          Agilent Whole Chr array_18 19 21 A_17_P32014191 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32014196 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11354576 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11354588 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17231177 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11354589 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32014192 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32014187 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32014194 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32014190 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11354575 - -
          Agilent Whole Chr array_18 19 21 A_17_P11354584 - -
          Agilent Whole Chr array_18 19 21 A_17_P32014197 - -
          Agilent Whole Chr array_18 19 21 A_17_P11354580 - -
          Agilent Whole Chr array_18 19 21 A_17_P11354581 - -
          Agilent Whole Chr array_18 19 21 A_17_P32014191 - -
          Agilent Whole Chr array_18 19 21 A_17_P32014193 - -
          Agilent Whole Chr array_18 19 21 A_17_P32014187 - -
          Agilent Whole Chr array_18 19 21 A_17_P11354576 - -
          Agilent Whole Chr array_18 19 21 A_17_P32014203 - -
          Agilent Whole Chr array_18 19 21 A_17_P11354588 - -
          Agilent Whole Chr array_18 19 21 A_17_P32014190 - -
          Agilent Whole Chr array_18 19 21 A_17_P11354587 - -
          Agilent Whole Chr array_18 19 21 A_17_P11354589 - -
          Agilent Whole Chr array_18 19 21 A_17_P32014194 - -
          Agilent Whole Chr array_18 19 21 A_17_P17231176 - -
          Agilent Whole Chr array_18 19 21 A_17_P17231175 - -
          Agilent Whole Chr array_18 19 21 A_17_P32014201 - -
          Agilent Whole Chr array_18 19 21 A_17_P11354583 - -
          Agilent Whole Chr array_18 19 21 A_17_P11354575 - -
          Agilent Whole Chr array_18 19 21 A_17_P32014196 - -
          Agilent Whole Chr array_18 19 21 A_17_P32014205 - -
          Agilent Whole Chr array_18 19 21 A_17_P11354590 - -
          Agilent Whole Chr array_18 19 21 A_17_P17231178 - -
          Agilent Whole Chr array_18 19 21 A_17_P17231177 - -
          Agilent Whole Chr array_18 19 21 A_17_P32014192 - -
          Agilent Whole Chr array_18 19 21 A_17_P11354577 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11354588 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32014190 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17231177 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11354575 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32014196 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32014194 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32014187 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32014192 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11354576 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11354589 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P29023 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P29029 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P110471 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 43755 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P110471 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11354575 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11354579 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11354546 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11354588 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11354585 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11354576 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11354581 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11354574 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11354590 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11354577 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17231178 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11354594 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11354589 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11354591 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P110471 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8068201 - -
          Affymetrix Human Gene 1.1 ST Array 8068200 - -
          Affymetrix HT HG-U133+ PM Array Plate 227424_PM_x_at - -
          Human Genome U133 Plus 2.0 Array 227424_x_at - -
          Human Genome U133 Plus 2.0 Array 227424_PM_x_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227424_x_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227424_x_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227424_x_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227424_x_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227424_x_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227424_x_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227424_x_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227424_x_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227424_x_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227424_x_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227424_x_at7 - -
          Human Genome U95 Set 53179_at - -
          Human Genome U95 Set 53179_PM_at - -
          Human Genome U133 Set 227424_x_at - -
          Human Genome U133 Set 227424_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A019159 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A019159 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 147385 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1728403 NR_026845.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1728403 AI369236.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1728403 AK026376.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1728403 BC007928.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1728403 NR_026845.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1728403 AI369236.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1728403 AK026376.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1728403 BC007928.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1728403 NR_026845.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1728403 AI369236.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1728403 AK026376.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1728403 BC007928.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1728403 NR_026845.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1728403 AI369236.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1728403 AK026376.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1728403 BC007928.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1728403 NR_026845.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1728403 AI369236.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1728403 AK026376.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1728403 BC007928.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1728403 NR_026845.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1728403 AI369236.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1728403 AK026376.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1728403 BC007928.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1728403 NR_026845.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1728403 AI369236.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1728403 AK026376.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1728403 BC007928.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1728403 NR_026845.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1728403 AI369236.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1728403 AK026376.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1728403 BC007928.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1728403 NR_026845.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1728403 AI369236.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1728403 AK026376.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1728403 BC007928.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline