Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00918261_s1
          See other LOC729324 GE Assays ›
          Gene Symbol:
          LOC729324
          Gene Name:
          hCG1986447
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00918261_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC729324
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC041895.1 - 1 244 85
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BI561503.1 - 1 248 Chr. 2 - 65227326 - 65227945 [-] on Build GRCh38 85
          GenBankmRNA BI668545.1 - 1 340 Chr. 2 - 65227385 - 65228036 [-] on Build GRCh38 85
          GenBankmRNA HY031307.1 - 1 269 Chr. 2 - 65227512 - 65227970 [-] on Build GRCh38 85
          GenBankmRNA DB451575.1 - 1 280 Chr. 2 - 65227488 - 65227981 [-] on Build GRCh38 85

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ARP2 actin related protein 2 homolog ACTR2 ARP2 Anti-sense Chr. 2 - 65227695 - 65271253 on NCBI Build GRCh38
          Downstream sprouty related EVH1 domain containing 2 SPRED2 Spred-2 79.4kb Sense Chr. 2 - 65307425 - 65432599 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 632493 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3298440 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 30543 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3298440 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P35709673 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P35709673 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000377982.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000377982.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000377982.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000377982.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000377982.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000377982.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000377982.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000377982.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000377982.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000377982.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000377982.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000377982.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000471552.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000471552.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000471552.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000471552.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000471552.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000476840.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000476840.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000476840.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000476840.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000476840.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000476840.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000542850.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000542850.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000542850.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000542850.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000542850.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000542850.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000542850.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000542850.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 ENST00000542850.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000260641.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000377982.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000377982.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000377982.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000377982.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000377982.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000377982.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000377982.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000377982.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000377982.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000377982.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000377982.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000377982.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000471552.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000471552.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000471552.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000471552.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000471552.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000476840.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000476840.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000476840.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000476840.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000476840.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000476840.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000542850.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000542850.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000542850.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000542850.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000542850.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000542850.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000542850.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000542850.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 ENST00000542850.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_001005386.2 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_005722.3 Refseq
          Affymetrix Human Clariom D Assay TC0200007835.hg.1 NM_005722.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 231980 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3301468 BC041895.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3224888 AK055581.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3224888 AK055581.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3301468 BC041895.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0