Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019568.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019568.hg.1 |
NM_181539.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019568.hg.1 |
TC17001470.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010637.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010637.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010637.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010637.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010637.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010637.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010637.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010637.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010637.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010637.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010637.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010637.hg.1 |
NM_181539.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010637.hg.1 |
NM_181539.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010637.hg.1 |
NM_181539.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010637.hg.1 |
NM_181539.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010637.hg.1 |
NM_181539.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010637.hg.1 |
NM_181539.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010637.hg.1 |
NM_181539.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010637.hg.1 |
NM_181539.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010637.hg.1 |
NM_181539.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010637.hg.1 |
NM_181539.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010637.hg.1 |
NM_181539.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183979.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183979.hg.1 |
NM_181539.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183979.hg.1 |
TC1700010637.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700183979.hg.1 |
NM_181539.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183979.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010637.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010637.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010637.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010637.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010637.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010637.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010637.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010637.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010637.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010637.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010637.hg.1 |
ENST00000335552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010637.hg.1 |
NM_181539.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010637.hg.1 |
NM_181539.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010637.hg.1 |
NM_181539.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010637.hg.1 |
NM_181539.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010637.hg.1 |
NM_181539.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010637.hg.1 |
NM_181539.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010637.hg.1 |
NM_181539.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010637.hg.1 |
NM_181539.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010637.hg.1 |
NM_181539.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010637.hg.1 |
NM_181539.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010637.hg.1 |
NM_181539.4 |
Refseq |