Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00920494_m1
          See other TBP GE Assays ›
          Gene Symbol:
          TBP
          Gene Name:
          TATA-box binding protein
          Gene Aliases:
          GTF2D, GTF2D1, HDL4, SCA17, TFIID
          Chromosome Location:
          Chr.6: 170554333 - 170572870 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00920494_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TBP
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003194.4 NP_003185.1 2 - 3 330 104
          GenBank mRNA AK298272.1 - 2 - 3 281 104
          AK315222.1 - 2 - 3 296 104
          BC109053.1 - 2 - 3 244 40032492 104
          BC109054.1 - 2 - 3 248 40032493 104
          BC110341.1 - 2 - 3 292 5761426 104
          BM720195.1 - 2 - 3 330 104
          BT019657.1 - 1 - 2 51 104
          CR456776.1 - 1 - 2 51 104
          FJ770048.1 - 1 - 2 132 104
          M34960.1 - 1 - 2 51 104
          M55654.1 - 2 - 3 292 104
          X54993.1 - 1 - 2 149 104
          Z22828.1 - 2 - 3 289 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600010343.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06001201.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TBP

          Panther Classification:

          Molecular Function -

          general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription initiation from RNA polymerase I promoter
          transcription elongation from RNA polymerase I promoter
          termination of RNA polymerase I transcription
          transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          transcription elongation from RNA polymerase II promoter
          transcription from RNA polymerase III promoter
          spermatogenesis
          viral process
          snRNA transcription from RNA polymerase II promoter
          positive regulation of gene expression, epigenetic
          positive regulation of transcription, DNA-templated
          regulation of signal transduction by p53 class mediator
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          RNA polymerase II repressing transcription factor binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          enzyme binding
          transcription regulatory region DNA binding
          repressing transcription factor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000230354.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000230354.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000230354.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000230354.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000230354.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000230354.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000230354.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000230354.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000230354.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000230354.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000230354.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000230354.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000230354.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000230354.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000230354.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000230354.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000230354.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000230354.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000230354.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000230354.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000392092.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000392092.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000392092.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000392092.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000392092.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000392092.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000392092.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000392092.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000392092.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000392092.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000392092.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000392092.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000392092.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000392092.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000392092.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000392092.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000392092.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000392092.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000392092.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000392092.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000392092.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000421512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000421512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000421512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000421512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000421512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000421512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000421512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000421512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000421512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000421512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000421512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000421512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000421512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000421512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000421512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000423353.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000423353.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000423353.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000423353.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000423353.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000423353.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000423353.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000423353.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000423353.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000446829.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000446829.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000446829.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000446829.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000446829.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000540980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000540980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000540980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000540980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000540980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000540980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000540980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000540980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000540980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000540980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000540980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000540980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000540980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000540980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000540980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000540980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000540980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000540980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000540980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000616883.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000616883.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000616883.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000616883.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000616883.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000616883.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000616883.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000616883.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000616883.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000616883.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000616883.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000616883.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000616883.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000616883.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000616883.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000616883.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000616883.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000616883.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 ENST00000616883.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_001172085.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_001172085.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_001172085.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_001172085.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_001172085.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_001172085.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_001172085.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_001172085.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_001172085.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_001172085.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_001172085.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_001172085.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_001172085.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_001172085.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_001172085.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_001172085.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_001172085.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_001172085.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_001172085.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_003194.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_003194.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_003194.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_003194.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_003194.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_003194.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_003194.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_003194.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_003194.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_003194.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_003194.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_003194.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_003194.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_003194.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_003194.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_003194.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_003194.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_003194.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_003194.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_003194.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010343.hg.1 NM_003194.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0