Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00921017_s1
          See other LINC00889 GE Assays ›
          Gene Symbol:
          LINC00889
          Gene Name:
          long intergenic non-protein coding RNA 889
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 138614731 - 138617638 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00921017_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00889
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026935.1 1 2095 79
          GenBank mRNA AK055234.1 - 1 1644 79
          AK055382.1 - 1 1779 79

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream Zic family member 3 ZIC3 HTX,
          HTX1,
          VACTERLX,
          ZNF203
          -1.0mb Anti-sense Chr. X - 137566187 - 137577691 on NCBI Build GRCh38
          Downstream fibroblast growth factor 13 FGF13 FGF-13,
          FGF2,
          FHF-2,
          FHF2
          14.3kb Sense Chr. X - 138631573 - 139222889 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32716974 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32716977 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32716972 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32716975 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32716978 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32716979 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32716976 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 93526 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 24241 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P360268 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P360268 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 35031 - -
          Agilent Human Genome CGH Microarray 44B A_14_P137194 - -
          Agilent Human Genome CGH Microarray 44A A_14_P137194 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 20934 - -
          Agilent Human Genome CGH + SNP Microarray A_14_P137194 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P360268 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P360268 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32716984 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32716979 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32716983 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32716978 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17403315 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32716980 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32716981 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32716975 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32716976 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32716985 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32716977 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17403315 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32716977 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32716983 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32716985 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32716976 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32716980 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32716975 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32716979 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32716981 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32716984 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32716978 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P360268 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P137194 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P137194 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P137194 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P360268 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 131541 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 65218 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P137194 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11983723 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11983729 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11983728 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11983725 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11983739 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17403314 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11983732 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11983721 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11983737 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11983735 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P137194 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8175443 - -
          Affymetrix Human Gene 1.1 ST Array 8175442 - -
          Affymetrix HT HG-U133+ PM Array Plate 1553357_PM_at - -
          Human Genome U133 Plus 2.0 Array 1553357_at - -
          Human Genome U133 Plus 2.0 Array 1553357_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553357_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553357_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553357_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553357_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553357_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553357_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553357_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553357_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553357_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553357_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553357_at4 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A040467 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A040467 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0