Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000091.hg.1 |
ENST00000312165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000091.hg.1 |
ENST00000534750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000091.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000091.hg.1 |
TC11000007.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
ENST00000312165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
ENST00000312165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
ENST00000312165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
ENST00000312165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
ENST00000312165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
ENST00000312165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
ENST00000312165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
ENST00000312165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
ENST00000527946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
ENST00000527946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
ENST00000527946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
ENST00000534750.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
ENST00000534750.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
ENST00000534750.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
ENST00000534750.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
ENST00000534750.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
ENST00000534750.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
ENST00000534750.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
ENST00000534750.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
ENST00000534750.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
ENST00000534750.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
NM_001276700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
NM_001276700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
NM_001276700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
NM_001276700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
NM_001276700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
NM_001276700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
NM_001276700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
NM_001276700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
NM_001276700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
NM_001276700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006442.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000092.hg.1 |
ENST00000312165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000092.hg.1 |
ENST00000534750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000092.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000092.hg.1 |
TC11000007.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100144207.hg.1 |
ENST00000312165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144207.hg.1 |
ENST00000527946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144207.hg.1 |
ENST00000534750.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144207.hg.1 |
NM_001276700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144207.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144207.hg.1 |
TC1100006442.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100144207.hg.1 |
NM_001276700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144207.hg.1 |
ENST00000312165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144207.hg.1 |
ENST00000527946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144207.hg.1 |
ENST00000534750.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144207.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144205.hg.1 |
ENST00000312165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144205.hg.1 |
ENST00000534750.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144205.hg.1 |
NM_001276700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144205.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144205.hg.1 |
TC1100006442.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100144205.hg.1 |
ENST00000312165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144205.hg.1 |
ENST00000534750.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144205.hg.1 |
NM_001276700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144205.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
ENST00000312165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
ENST00000312165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
ENST00000312165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
ENST00000312165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
ENST00000312165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
ENST00000312165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
ENST00000312165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
ENST00000312165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
ENST00000527946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
ENST00000527946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
ENST00000527946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
ENST00000534750.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
ENST00000534750.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
ENST00000534750.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
ENST00000534750.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
ENST00000534750.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
ENST00000534750.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
ENST00000534750.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
ENST00000534750.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
ENST00000534750.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
ENST00000534750.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
NM_001276700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
NM_001276700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
NM_001276700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
NM_001276700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
NM_001276700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
NM_001276700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
NM_001276700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
NM_001276700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
NM_001276700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
NM_001276700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
NM_138329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006442.hg.1 |
NM_138329.2 |
Refseq |