Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00921550_g1
          See other PPP1R15A GE Assays ›
          Gene Symbol:
          PPP1R15A
          Gene Name:
          protein phosphatase 1 regulatory subunit 15A
          Gene Aliases:
          GADD34
          Chromosome Location:
          Chr.19: 48872392 - 48876062 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00921550_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PPP1R15A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_014330.3 NP_055145.3 1 - 2 144
          GenBank mRNA AK001361.1 - 1 - 2 235 144
          AK225632.1 - 1 - 2 144
          BC003067.2 - 1 - 2 3507556 144
          DA484808.1 - 1 - 2 144
          U83981.1 - 1 - 2 144

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900008496.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19000711.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK001361.1, BC003067.2, NM_014330.3, U83981.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PPP1R15A

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          apoptotic process
          cellular response to DNA damage stimulus
          aging
          negative regulation of cell population proliferation
          negative regulation of peptidyl-threonine phosphorylation
          negative regulation of protein processing
          endoplasmic reticulum unfolded protein response
          positive regulation of translational initiation in response to stress
          negative regulation of phosphoprotein phosphatase activity
          positive regulation of phosphoprotein phosphatase activity
          cellular response to UV
          response to endoplasmic reticulum stress
          negative regulation of protein dephosphorylation
          response to immobilization stress
          regulation of translational initiation by eIF2 alpha dephosphorylation
          response to cocaine
          positive regulation of apoptotic process
          regulation of angiogenesis
          negative regulation of cell death
          regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          peptidyl-serine dephosphorylation
          protein localization to endoplasmic reticulum
          cellular response to antibiotic
          cellular response to carbohydrate stimulus
          cellular response to xenobiotic stimulus
          cellular response to methyl methanesulfonate
          cellular response to calcium ion starvation
          response to environmental enrichment
          cellular response to oxygen-glucose deprivation
          positive regulation of peptidyl-serine dephosphorylation
          negative regulation of PERK-mediated unfolded protein response
          negative regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation
          positive regulation of endoplasmic reticulum stress-induced eIF2 alpha dephosphorylation
          cellular response to desipramine
          cellular response to cordycepin
          cellular response to gold(3+)
          cellular response to methamphetamine hydrochloride
          negative regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
          protein binding
          protein phosphatase 1 binding
          protein phosphatase regulator activity
          protein kinase binding
          protein phosphatase activator activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR19009825.hg.1 ENST00000200453.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19009825.hg.1 NM_014330.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19009825.hg.1 TC19000711.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19009829.hg.1 ENST00000200453.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19009829.hg.1 ENST00000540695.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19009829.hg.1 ENST00000544084.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19009829.hg.1 NM_014330.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19009829.hg.1 TC19000711.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19009831.hg.1 ENST00000200453.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19009831.hg.1 NM_014330.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19009831.hg.1 TC19000711.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19009834.hg.1 ENST00000200453.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19009834.hg.1 ENST00000540695.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19009834.hg.1 ENST00000544084.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19009834.hg.1 NM_014330.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19009834.hg.1 TC19000711.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1900008496.hg.1 ENST00000200453.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008496.hg.1 ENST00000200453.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008496.hg.1 ENST00000200453.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008496.hg.1 ENST00000200453.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008496.hg.1 ENST00000200453.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008496.hg.1 ENST00000200453.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008496.hg.1 ENST00000600406.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008496.hg.1 ENST00000600406.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008496.hg.1 ENST00000600406.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008496.hg.1 ENST00000600406.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008496.hg.1 NM_014330.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008496.hg.1 NM_014330.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008496.hg.1 NM_014330.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008496.hg.1 NM_014330.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008496.hg.1 NM_014330.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008496.hg.1 NM_014330.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19009832.hg.1 ENST00000200453.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19009832.hg.1 ENST00000544084.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19009832.hg.1 NM_014330.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19009832.hg.1 TC19000711.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19009828.hg.1 ENST00000200453.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19009828.hg.1 ENST00000544084.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19009828.hg.1 NM_014330.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19009828.hg.1 TC19000711.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19009830.hg.1 ENST00000200453.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19009830.hg.1 ENST00000544084.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19009830.hg.1 NM_014330.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19009830.hg.1 TC19000711.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1900165385.hg.1 ENST00000200453.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900165385.hg.1 NM_014330.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900165385.hg.1 TC1900008496.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900165385.hg.1 NM_014330.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900165385.hg.1 ENST00000200453.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900165386.hg.1 ENST00000200453.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900165386.hg.1 NM_014330.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900165386.hg.1 TC1900008496.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900165386.hg.1 ENST00000200453.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900165386.hg.1 NM_014330.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900165388.hg.1 ENST00000200453.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900165388.hg.1 NM_014330.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900165388.hg.1 TC1900008496.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900165388.hg.1 ENST00000200453.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900165388.hg.1 NM_014330.3 Refseq
          Affymetrix Human Clariom D Assay TC1900008496.hg.1 ENST00000200453.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900008496.hg.1 ENST00000200453.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900008496.hg.1 ENST00000200453.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900008496.hg.1 ENST00000200453.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900008496.hg.1 ENST00000200453.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900008496.hg.1 ENST00000200453.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900008496.hg.1 ENST00000600406.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008496.hg.1 ENST00000600406.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008496.hg.1 ENST00000600406.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008496.hg.1 ENST00000600406.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008496.hg.1 NM_014330.3 Refseq
          Affymetrix Human Clariom D Assay TC1900008496.hg.1 NM_014330.3 Refseq
          Affymetrix Human Clariom D Assay TC1900008496.hg.1 NM_014330.3 Refseq
          Affymetrix Human Clariom D Assay TC1900008496.hg.1 NM_014330.3 Refseq
          Affymetrix Human Clariom D Assay TC1900008496.hg.1 NM_014330.3 Refseq
          Affymetrix Human Clariom D Assay TC1900008496.hg.1 NM_014330.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 143553 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix