Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00922471_g1
          See other PDSS1 GE Assays ›
          Gene Symbol:
          PDSS1
          Gene Name:
          prenyl (decaprenyl) diphosphate synthase, subunit 1
          Gene Aliases:
          COQ1, COQ10D2, DPS, SPS, TPRT, TPT, TPT 1, hDPS1
          Chromosome Location:
          Chr.10: 26697052 - 26746798 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00922471_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PDSS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001321978.1 NP_001308907.1 9 - 10 1202 67
          NM_001321979.1 NP_001308908.1 11 - 12 1476 67
          NM_014317.4 NP_055132.2 11 - 12 1393 67
          XM_011519437.2 XP_011517739.1 8 - 9 1793 67
          XM_017016011.1 XP_016871500.1 9 - 10 871 67
          GenBank mRNA AB209763.1 - 10 - 11 3435 67
          AB210838.1 - 11 - 12 1151 67
          AF118395.1 - 10 - 11 1013 67
          AK024802.1 - 11 - 12 1158 67
          AK223414.1 - 11 - 12 1158 67
          AK296288.1 - 4 - 5 463 67
          BC049211.1 - 11 - 12 1103 67
          BC063635.1 - 9 - 10 930 4397753 67

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PDSS1

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquinone biosynthetic process
          isoprenoid biosynthetic process
          protein heterotetramerization
          trans-hexaprenyltranstransferase activity
          metal ion binding
          protein heterodimerization activity
          trans-octaprenyltranstransferase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000449300.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000449300.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000449300.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000458171.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000458171.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000470978.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000470978.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000470978.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000470978.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000470978.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000470978.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000470978.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000473224.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000473224.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000473224.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000473224.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000473224.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000491711.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000491711.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000491711.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000491711.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000491711.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000491711.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000491711.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 ENST00000491711.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000376215.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000449300.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000449300.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000449300.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000458171.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000458171.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000470978.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000470978.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000470978.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000470978.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000470978.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000470978.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000470978.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000473224.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000473224.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000473224.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000473224.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000473224.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000491711.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000491711.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000491711.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000491711.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000491711.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000491711.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000491711.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 ENST00000491711.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 NM_014317.4 Refseq
          Affymetrix Human Clariom D Assay TC1000007081.hg.1 NM_014317.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0