Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000272452.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000272452.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000272452.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000272452.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000272452.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000272452.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000272452.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000272452.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000272452.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000272452.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000272452.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000272452.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000409309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000409309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000409309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000409309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000409309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000409309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000409309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000494122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000494122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000494122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000494122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000494122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000494122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000494122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000494122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
ENST00000494122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
NM_006588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
NM_006588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
NM_006588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
NM_006588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
NM_006588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
NM_006588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
NM_006588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
NM_006588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
NM_006588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
NM_006588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
NM_006588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008801.hg.1 |
NM_006588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000272452.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000272452.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000272452.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000272452.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000272452.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000272452.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000272452.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000272452.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000272452.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000272452.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000272452.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000272452.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000409309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000409309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000409309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000409309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000409309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000409309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000409309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000494122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000494122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000494122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000494122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000494122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000494122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000494122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000494122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
ENST00000494122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
NM_006588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
NM_006588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
NM_006588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
NM_006588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
NM_006588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
NM_006588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
NM_006588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
NM_006588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
NM_006588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
NM_006588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
NM_006588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008801.hg.1 |
NM_006588.3 |
Refseq |