Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00924629_m1
          See other FLNA GE Assays ›
          Gene Symbol:
          FLNA
          Gene Name:
          filamin A
          Gene Aliases:
          ABP-280, ABPX, CSBS, CVD1, FLN, FLN-A, FLN1, FMD, MNS, NHBP, OPD, OPD1, OPD2, XLVD, XMVD
          Chromosome Location:
          Chr.X: 154348529 - 154374638 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00924629_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FLNA
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001456.3 NP_001447.2 29 - 30 5199 116
          XM_011531128.1 XP_011529430.1 29 - 30 5194 116
          XM_011531130.1 XP_011529432.1 29 - 30 5194 116
          GenBank mRNA AB191259.1 - 28 - 29 4922 116
          AB371575.1 - 28 - 29 4922 116
          AB371578.1 - 28 - 29 4922 116
          AB371579.1 - 28 - 29 4922 116
          AK074048.1 - 8 - 9 1399 116
          AK090427.2 - 28 - 29 4988 116
          AK304255.1 - 2 - 3 234 116
          AL157419.1 - 6 - 7 963 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00011194.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X001530.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK304255.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FLNA

          Gene Ontology Categories:

          Function(s) Process(es)

          platelet degranulation
          adenylate cyclase-inhibiting dopamine receptor signaling pathway
          negative regulation of transcription by RNA polymerase I
          platelet activation
          regulation of cell migration
          actin cytoskeleton reorganization
          cell junction assembly
          protein localization to cell surface
          negative regulation of protein catabolic process
          mRNA transcription by RNA polymerase II
          negative regulation of apoptotic process
          receptor clustering
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of DNA-binding transcription factor activity
          wound healing, spreading of cells
          establishment of protein localization
          protein stabilization
          cytoplasmic sequestering of protein
          actin crosslink formation
          cilium assembly
          platelet aggregation
          semaphorin-plexin signaling pathway
          mitotic spindle assembly
          positive regulation of substrate adhesion-dependent cell spreading
          positive regulation of integrin-mediated signaling pathway
          G protein-coupled receptor binding
          RNA binding
          protein binding
          transcription factor binding
          kinase binding
          small GTPase binding
          mu-type opioid receptor binding
          Fc-gamma receptor I complex binding
          protein homodimerization activity
          SMAD binding
          actin filament binding
          GTPase binding
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR0X020397.hg.1 ENST00000344736.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X020397.hg.1 ENST00000360319.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X020397.hg.1 ENST00000369850.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X020397.hg.1 ENST00000369852.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X020397.hg.1 ENST00000369856.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X020397.hg.1 ENST00000422373.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X020397.hg.1 NM_001110556.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X020397.hg.1 NM_001456.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X020397.hg.1 TC0X001530.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0X00176394.hg.1 ENST00000344736.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00176394.hg.1 ENST00000360319.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00176394.hg.1 ENST00000369850.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00176394.hg.1 ENST00000369856.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00176394.hg.1 ENST00000420627.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00176394.hg.1 ENST00000422373.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00176394.hg.1 ENST00000490936.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00176394.hg.1 ENST00000610817.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00176394.hg.1 NM_001110556.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00176394.hg.1 NM_001456.3 Refseq
          Affymetrix Human Clariom D Assay PSR0X00176394.hg.1 TC0X00011194.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00176394.hg.1 ENST00000344736.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00176394.hg.1 ENST00000360319.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00176394.hg.1 ENST00000369850.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00176394.hg.1 ENST00000369856.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00176394.hg.1 ENST00000420627.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00176394.hg.1 ENST00000422373.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00176394.hg.1 ENST00000490936.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00176394.hg.1 ENST00000610817.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00176394.hg.1 NM_001110556.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00176394.hg.1 NM_001456.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0