Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010141.hg.1 |
TC22000565.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010141.hg.1 |
ENST00000417194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010141.hg.1 |
NR_024448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000417194.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000421064.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000421064.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000422506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000422506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000423913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000423913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000437862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000445682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000445682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000451837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000451837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000451919.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000452737.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000452737.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000455485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000455485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000458554.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000608454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
NR_024448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000423913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
TC2200009326.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000417194.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000421064.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000421064.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000422506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000422506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000423913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000437862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000445682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000445682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000451837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000451837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000451919.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000452737.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000452737.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000455485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000455485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000458554.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
ENST00000608454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169888.hg.1 |
NR_024448.2 |
Refseq |