Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00924808_m1
          See other PTGS1 GE Assays ›
          Gene Symbol:
          PTGS1
          Gene Name:
          prostaglandin-endoperoxide synthase 1
          Gene Aliases:
          COX1, COX3, PCOX1, PES-1, PGG/HS, PGHS-1, PGHS1, PHS1, PTGHS
          Chromosome Location:
          Chr.9: 122369906 - 122395703 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00924808_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PTGS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000962.3 NP_000953.2 5 - 6 574 72
          NM_001271165.1 NP_001258094.1 4 - 5 523 72
          NM_001271166.1 NP_001258095.1 5 - 6 800 72
          NM_001271367.1 NP_001258296.1 5 - 6 545 72
          NM_001271368.1 NP_001258297.1 6 - 7 1022 72
          NM_080591.2 NP_542158.1 5 - 6 574 72
          XM_005252105.3 XP_005252162.1 6 - 7 749 72
          XM_011518875.2 XP_011517177.1 6 - 7 1010 72
          XM_011518876.2 XP_011517178.1 6 - 7 4620 72
          XM_017014929.1 XP_016870418.1 8 - 9 984 72
          GenBank mRNA AK290022.1 - 5 - 6 527 72
          AK295221.1 - 6 - 7 1007 72
          AK304403.1 - 4 - 5 523 72
          AK308222.1 - 5 - 6 800 72
          BC029840.1 - 5 - 6 546 5174923 72
          CD014049.1 - 5 - 6 511 72
          DQ180741.1 - 4 - 5 587 72
          DQ180742.1 - 4 - 5 587 72
          M59979.1 - 4 - 5 499 72
          S36219.1 - 4 - 5 494 72
          S36271.1 - 4 - 5 494 72
          S78220.1 - 4 - 5 494 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900008660.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09000614.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PTGS1

          Gene Ontology Categories:

          Function(s) Process(es)

          prostaglandin biosynthetic process
          lipid metabolic process
          xenobiotic metabolic process
          inflammatory response
          response to oxidative stress
          aging
          learning
          memory
          regulation of blood pressure
          response to organonitrogen compound
          negative regulation of norepinephrine secretion
          sensory perception of pain
          cyclooxygenase pathway
          negative regulation of epinephrine secretion
          maintenance of blood-brain barrier
          regulation of cell population proliferation
          positive regulation of vasoconstriction
          positive regulation of smooth muscle contraction
          response to corticosterone
          response to fatty acid
          cellular oxidant detoxification
          peroxidase activity
          prostaglandin-endoperoxide synthase activity
          lipid binding
          heme binding
          metal ion binding
          dioxygenase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR09007099.hg.1 ENST00000223423.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09007099.hg.1 ENST00000362012.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09007099.hg.1 ENST00000373698.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09007099.hg.1 ENST00000540753.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09007099.hg.1 NM_000962.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09007099.hg.1 NM_080591.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09007099.hg.1 TC09000614.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09007100.hg.1 ENST00000223423.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09007100.hg.1 ENST00000362012.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09007100.hg.1 ENST00000373698.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09007100.hg.1 ENST00000540753.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09007100.hg.1 NM_000962.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09007100.hg.1 NM_080591.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09007100.hg.1 TC09000614.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0900157872.hg.1 ENST00000223423.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0900157872.hg.1 ENST00000362012.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900157872.hg.1 ENST00000373698.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900157872.hg.1 ENST00000540753.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900157872.hg.1 ENST00000619306.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900157872.hg.1 NM_000962.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900157872.hg.1 NM_001271165.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900157872.hg.1 NM_001271166.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900157872.hg.1 NM_001271367.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900157872.hg.1 NM_001271368.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900157872.hg.1 NM_080591.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900157872.hg.1 TC0900008660.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900157872.hg.1 ENST00000223423.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0900157872.hg.1 ENST00000362012.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900157872.hg.1 ENST00000373698.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900157872.hg.1 ENST00000540753.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900157872.hg.1 ENST00000619306.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900157872.hg.1 NM_000962.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900157872.hg.1 NM_001271165.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900157872.hg.1 NM_001271166.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900157872.hg.1 NM_001271367.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900157872.hg.1 NM_001271368.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900157872.hg.1 NM_080591.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 ENST00000223423.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 ENST00000362012.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 ENST00000373698.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 ENST00000540753.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 ENST00000614910.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 ENST00000619306.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 NM_000962.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 NM_001271164.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 NM_001271165.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 NM_001271166.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 NM_001271367.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 NM_001271368.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 NM_080591.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 TC0900008660.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 ENST00000223423.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 ENST00000362012.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 ENST00000373698.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 ENST00000540753.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 ENST00000614910.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 ENST00000619306.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 NM_000962.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 NM_001271164.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 NM_001271165.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 NM_001271166.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 NM_001271367.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 NM_001271368.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900157873.hg.1 NM_080591.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0