Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00925202_g1
          See other PRKCA GE Assays ›
          Gene Symbol:
          PRKCA
          Gene Name:
          protein kinase C alpha
          Gene Aliases:
          AAG6, PKC-alpha, PKCA, PRKACA
          Chromosome Location:
          Chr.17: 66302640 - 66810744 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00925202_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PRKCA
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002737.2 NP_002728.1 7 - 8 859 92
          XM_017024836.1 XP_016880325.1 7 - 8 1027 92
          XM_017024837.1 XP_016880326.1 6 - 7 690 92
          XM_017024838.1 XP_016880327.1 7 - 8 786 92
          XM_017024839.1 XP_016880328.1 6 - 7 1725 92
          XM_017024840.1 XP_016880329.1 7 - 8 768 92
          XM_017024841.1 XP_016880330.1 7 - 8 1027 92
          GenBank mRNA AB209475.1 - 8 - 9 868 92
          AB451258.1 - 7 - 8 815 92
          AB451383.1 - 7 - 8 815 92
          AY633609.1 - 7 - 8 820 92
          BC053321.1 - 7 - 8 1031 92
          BC109273.1 - 7 - 8 841 40028305 92
          BC109274.1 - 7 - 8 841 40028308 92
          M22199.1 - 7 - 8 774 92
          X52479.1 - 7 - 8 842 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700008661.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17000783.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PRKCA

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          positive regulation of endothelial cell proliferation
          desmosome assembly
          protein phosphorylation
          mitotic nuclear membrane disassembly
          cell adhesion
          activation of adenylate cyclase activity
          negative regulation of adenylate cyclase activity
          positive regulation of endothelial cell migration
          positive regulation of cardiac muscle hypertrophy
          peptidyl-serine phosphorylation
          platelet activation
          positive regulation of cell migration
          positive regulation of lipopolysaccharide-mediated signaling pathway
          negative regulation of glial cell apoptotic process
          histone H3-T6 phosphorylation
          intracellular signal transduction
          regulation of mRNA stability
          positive regulation of blood vessel endothelial cell migration
          positive regulation of macrophage differentiation
          positive regulation of angiogenesis
          positive regulation of cell adhesion
          positive regulation of mitotic cell cycle
          regulation of insulin secretion
          positive regulation of ERK1 and ERK2 cascade
          response to interleukin-1
          regulation of platelet aggregation
          apoptotic signaling pathway
          positive regulation of dense core granule biogenesis
          protein kinase activity
          protein serine/threonine kinase activity
          protein kinase C activity
          calcium-dependent protein kinase C activity
          protein binding
          ATP binding
          zinc ion binding
          enzyme binding
          histone kinase activity (H3-T6 specific)

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000583361.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000583361.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000583361.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000583361.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000583361.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000583775.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000583775.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 ENST00000583775.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17010214.hg.1 ENST00000284384.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17010214.hg.1 ENST00000413366.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17010214.hg.1 NM_002737.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17010214.hg.1 TC17000783.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1700164752.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1700164752.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1700164752.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700164752.hg.1 NM_002737.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700164752.hg.1 TC1700008661.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700164752.hg.1 NM_002737.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700164752.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1700164752.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1700164752.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000284384.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000413366.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000578063.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000583361.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000583361.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000583361.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000583361.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000583361.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000583775.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000583775.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 ENST00000583775.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 NM_002737.2 Refseq
          Affymetrix Human Clariom D Assay TC1700008661.hg.1 NM_002737.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0