Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00929941_m1
          See other C2CD5 GE Assays ›
          Gene Symbol:
          C2CD5
          Gene Name:
          C2 calcium dependent domain containing 5
          Gene Aliases:
          CDP138, KIAA0528
          Chromosome Location:
          Chr.12: 22448546 - 22544546 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00929941_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C2CD5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001286173.1 NP_001273102.1 25 - 26 2944 88
          NM_001286174.1 NP_001273103.1 23 - 24 3078 88
          NM_001286175.1 NP_001273104.1 25 - 26 2947 88
          NM_001286176.1 NP_001273105.1 24 - 25 2938 88
          NM_001286177.1 NP_001273106.1 24 - 25 2911 88
          XM_005253539.3 XP_005253596.1 16 - 17 1630 88
          XM_017020264.1 XP_016875753.1 28 - 29 3230 88
          XM_017020265.1 XP_016875754.1 28 - 29 3200 88
          XM_017020266.1 XP_016875755.1 27 - 28 3179 88
          XM_017020267.1 XP_016875756.1 26 - 27 3158 88
          XM_017020268.1 XP_016875757.1 25 - 26 3092 88
          XM_017020270.1 XP_016875759.1 27 - 28 3038 88
          XM_017020271.1 XP_016875760.1 27 - 28 3011 88
          XM_017020272.1 XP_016875761.1 25 - 26 2939 88
          XM_017020273.1 XP_016875762.1 24 - 25 2888 88
          XM_017020274.1 XP_016875763.1 24 - 25 2870 88
          XM_017020276.1 XP_016875765.1 24 - 25 2843 88
          XM_017020277.1 XP_016875766.1 23 - 24 2822 88
          XM_017020279.1 XP_016875768.1 20 - 21 1953 88
          GenBank mRNA AK299353.1 - 23 - 24 2785 88
          BC117143.1 - 24 - 25 2774 40125694 88
          BC143859.1 - 24 - 25 2801 9052375 88
          BC143860.1 - 24 - 25 2774 9052376 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200010119.hg.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          vesicle fusion
          positive regulation of glucose transmembrane transport
          positive regulation of vesicle fusion
          cellular response to insulin stimulus
          insulin receptor signaling pathway via phosphatidylinositol 3-kinase
          intracellular protein transmembrane transport
          protein localization to plasma membrane
          positive regulation of protein targeting to membrane
          calcium ion binding
          calcium-dependent phospholipid binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 ENST00000333957.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 ENST00000396028.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 ENST00000446597.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 ENST00000536386.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 ENST00000539615.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 ENST00000542676.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 ENST00000543888.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 ENST00000545552.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 NM_001286173.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 NM_001286174.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 NM_001286175.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 NM_001286176.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 NM_001286177.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 NM_014802.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 TC1200010119.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 ENST00000536386.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 ENST00000333957.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 ENST00000396028.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 ENST00000446597.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 ENST00000539615.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 ENST00000542676.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 ENST00000543888.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 ENST00000545552.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 NM_001286173.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 NM_001286174.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 NM_001286175.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 NM_001286176.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 NM_001286177.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200176537.hg.1 NM_014802.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0