Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00930002_g1
          See other AICDA GE Assays ›
          Gene Symbol:
          AICDA
          Gene Name:
          activation-induced cytidine deaminase
          Gene Aliases:
          AID, ARP2, CDA2, HEL-S-284, HIGM2
          Chromosome Location:
          Chr.12: 8602166 - 8612970 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00930002_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AICDA
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020661.2 NP_065712.1 3 - 4 506 114
          GenBank mRNA AB040431.1 - 3 - 4 503 114
          AF529815.1 - 2 - 3 427 114
          AF529816.1 - 2 - 3 427 114
          AF529818.1 - 2 - 3 427 114
          AF529819.1 - 2 - 3 427 114
          AF529820.1 - 2 - 3 427 114
          AF529821.1 - 2 - 3 427 114
          AF529822.1 - 2 - 3 427 114
          AF529823.1 - 2 - 3 427 114
          AF529824.1 - 2 - 3 427 114
          AF529825.1 - 2 - 3 427 114
          AF529826.1 - 2 - 3 427 114
          AF529827.1 - 2 - 3 427 114
          AJ577811.1 - 1 - 2 219 114
          AY748364.1 - 1 - 2 209 114
          BC006296.2 - 3 - 4 503 4054915 114
          BT007402.1 - 2 - 3 427 114
          BX402063.2 - 3 - 4 520 115
          DQ431660.1 - 2 - 3 421 114
          EU794587.1 - 3 - 4 503 114

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200009831.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12001180.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene AICDA

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          cytidine deamination
          somatic diversification of immunoglobulins
          somatic hypermutation of immunoglobulin genes
          B cell differentiation
          isotype switching
          cellular response to lipopolysaccharide
          DNA demethylation
          negative regulation of DNA methylation-dependent heterochromatin assembly
          cytidine deaminase activity
          protein binding
          zinc ion binding
          ubiquitin protein ligase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000229335.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000229335.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000229335.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000229335.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000229335.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000229335.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000229335.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000229335.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000229335.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000229335.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000537228.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000537228.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000537228.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000537228.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000537228.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000537228.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000543081.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000543081.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000543081.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000543081.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000544516.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000544516.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000545512.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000545512.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000545512.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000545512.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000545512.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000545576.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000545576.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000545576.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000545576.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000545576.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000545576.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 ENST00000545576.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 NM_020661.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 NM_020661.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 NM_020661.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 NM_020661.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 NM_020661.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 NM_020661.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 NM_020661.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 NM_020661.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 NM_020661.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200009831.hg.1 NM_020661.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12015654.hg.1 ENST00000229335.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12015654.hg.1 NM_020661.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12015654.hg.1 TC12001180.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12015653.hg.1 ENST00000229335.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12015653.hg.1 ENST00000537228.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12015653.hg.1 NM_020661.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12015653.hg.1 TC12001180.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1200173910.hg.1 ENST00000229335.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1200173910.hg.1 ENST00000537228.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200173910.hg.1 ENST00000545512.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200173910.hg.1 ENST00000545576.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200173910.hg.1 NM_020661.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200173910.hg.1 TC1200009831.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200173910.hg.1 ENST00000545576.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200173910.hg.1 ENST00000229335.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1200173910.hg.1 ENST00000537228.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200173910.hg.1 ENST00000545512.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200173910.hg.1 NM_020661.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200173911.hg.1 ENST00000229335.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1200173911.hg.1 ENST00000545512.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200173911.hg.1 ENST00000545576.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200173911.hg.1 NM_020661.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200173911.hg.1 TC1200009831.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200173911.hg.1 ENST00000229335.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1200173911.hg.1 ENST00000545512.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200173911.hg.1 ENST00000545576.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200173911.hg.1 NM_020661.3 Refseq
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000229335.10 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000229335.10 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000229335.10 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000229335.10 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000229335.10 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000229335.10 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000229335.10 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000229335.10 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000229335.10 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000229335.10 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000537228.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000537228.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000537228.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000537228.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000537228.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000537228.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000543081.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000543081.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000543081.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000543081.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000544516.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000544516.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000545512.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000545512.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000545512.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000545512.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000545512.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000545576.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000545576.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000545576.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000545576.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000545576.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000545576.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 ENST00000545576.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 NM_020661.3 Refseq
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 NM_020661.3 Refseq
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 NM_020661.3 Refseq
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 NM_020661.3 Refseq
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 NM_020661.3 Refseq
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 NM_020661.3 Refseq
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 NM_020661.3 Refseq
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 NM_020661.3 Refseq
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 NM_020661.3 Refseq
          Affymetrix Human Clariom D Assay TC1200009831.hg.1 NM_020661.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline