Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000505753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000505753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000511871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000511871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000511871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000511871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
ENST00000511871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024374.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024374.hg.1 |
ENST00000369208.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024374.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024374.hg.1 |
TC06001969.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0600187381.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187381.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187381.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187381.hg.1 |
TC0600012670.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600187381.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187381.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187381.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000262901.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000369208.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000505753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000505753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000511871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000511871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000511871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000511871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
ENST00000511871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012670.hg.1 |
NM_005068.2 |
Refseq |