Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00933415_s1
          See other LOC729866 GE Assays ›
          Gene Symbol:
          LOC729866
          Gene Name:
          uncharacterized LOC729866
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00933415_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC729866
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK057458.1 - 1 129 63
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DB023838.1 - 1 129 Chr. 1 - 157925349 - 157925898 [-] on Build GRCh38 63

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream CD5 molecule like CD5L AIM,
          API6,
          CT-2,
          PRO229,
          SP-ALPHA,
          Spalpha,
          hAIM
          -82.0kb Sense Chr. 1 - 157827071 - 157841844 on NCBI Build GRCh38
          Downstream uncharacterized LOC105371458 LOC105371458 22.6kb Anti-sense Chr. 1 - 157925967 - 157949071 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00534495 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593241 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00534476 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593247 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593277 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593278 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593261 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00534514 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00534496 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593282 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593259 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593283 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593236 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00534471 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00534465 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593300 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593299 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593265 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593295 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593262 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593279 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593255 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00534509 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593273 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00534464 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593294 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593293 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593264 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593292 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593290 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00534477 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593263 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593269 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00534501 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00534533 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00534512 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593250 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593237 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00534525 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593274 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593258 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00534527 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593256 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593260 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00534474 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593243 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593268 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00534469 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593276 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593280 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20593240 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00534519 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00534475 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00534480 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 614776 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3365815 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P359043 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 20280 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 57653 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P56213219 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3365815 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P359043 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P359043 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P30338594 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7921343 - -
          Affymetrix Human Gene 1.1 ST Array 7921342 - -
          Affymetrix HT HG-U133+ PM Array Plate 1564235_PM_at - -
          Human Genome U133 Plus 2.0 Array 1564235_at - -
          Human Genome U133 Plus 2.0 Array 1564235_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564235_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564235_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564235_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564235_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564235_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564235_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564235_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564235_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564235_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564235_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564235_at2 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 230895 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3226056 AK057458.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0