Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18001041.hg.1 |
ENST00000269216.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18001041.hg.1 |
NM_005257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18001041.hg.1 |
TC18000094.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
ENST00000269216.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
ENST00000269216.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
ENST00000269216.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
ENST00000269216.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
ENST00000269216.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
ENST00000269216.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
ENST00000269216.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
ENST00000269216.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
ENST00000269216.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
ENST00000269216.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
ENST00000581694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
ENST00000581694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
ENST00000581694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
ENST00000581694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
ENST00000581694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
ENST00000581694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
ENST00000581694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
ENST00000581694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
ENST00000581694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
NM_005257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
NM_005257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
NM_005257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
NM_005257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
NM_005257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
NM_005257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
NM_005257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
NM_005257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
NM_005257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006852.hg.1 |
NM_005257.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800146909.hg.1 |
ENST00000269216.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800146909.hg.1 |
ENST00000581694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800146909.hg.1 |
NM_005257.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800146909.hg.1 |
TC1800006852.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800146909.hg.1 |
ENST00000269216.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800146909.hg.1 |
ENST00000581694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800146909.hg.1 |
NM_005257.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
ENST00000269216.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
ENST00000269216.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
ENST00000269216.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
ENST00000269216.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
ENST00000269216.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
ENST00000269216.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
ENST00000269216.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
ENST00000269216.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
ENST00000269216.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
ENST00000269216.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
ENST00000581694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
ENST00000581694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
ENST00000581694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
ENST00000581694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
ENST00000581694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
ENST00000581694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
ENST00000581694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
ENST00000581694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
ENST00000581694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
NM_005257.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
NM_005257.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
NM_005257.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
NM_005257.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
NM_005257.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
NM_005257.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
NM_005257.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
NM_005257.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
NM_005257.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006852.hg.1 |
NM_005257.5 |
Refseq |