Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003452.hg.1 |
ENST00000525748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003452.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003452.hg.1 |
TC11000258.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003454.hg.1 |
ENST00000525748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003454.hg.1 |
ENST00000528440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003454.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003454.hg.1 |
TC11000258.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000528440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000528440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000528440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000528440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000528440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000528440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000528440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000528440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
ENST00000528440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003451.hg.1 |
ENST00000525748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003451.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003451.hg.1 |
TC11000258.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000528440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000528440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000528440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000528440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000528440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000528440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000528440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000528440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
ENST00000528440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007055.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149741.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149741.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149741.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149741.hg.1 |
TC1100007055.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100149741.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149741.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149741.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149744.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149744.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149744.hg.1 |
ENST00000528440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149744.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149744.hg.1 |
TC1100007055.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100149744.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149744.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149744.hg.1 |
ENST00000528440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149744.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149742.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149742.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149742.hg.1 |
NM_004211.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149742.hg.1 |
TC1100007055.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100149742.hg.1 |
ENST00000298923.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149742.hg.1 |
ENST00000525748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149742.hg.1 |
NM_004211.4 |
Refseq |