Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs00938325_m1
See other PDE4D GE Assays ›
Gene Symbol:
PDE4D
Gene Name:
phosphodiesterase 4D
Gene Aliases:
ACRDYS2, DPDE3, HSPDE4D, PDE43, PDE4DN2, STRK1
Chromosome Location:
Chr.5: 58969038 - 60488098 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs00938325_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: PDE4D
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001104631.1
NP_001098101.1
6 - 7
1086
62
NM_001165899.1
NP_001159371.1
8 - 9
977
62
NM_001197218.1
NP_001184147.1
6 - 7
825
62
NM_001197219.1
NP_001184148.1
6 - 7
603
62
NM_001197220.1
NP_001184149.1
6 - 7
1241
62
NM_001197221.1
NP_001184150.1
2 - 3
561
62
NM_001197222.1
NP_001184151.1
1 - 2
647
62
NM_006203.4
NP_006194.2
6 - 7
629
62
XM_005248537.3
XP_005248594.1
6 - 7
1783
62
XM_005248538.4
XP_005248595.1
7 - 8
3684
62
XM_011543469.1
XP_011541771.1
9 - 10
1066
62
XM_011543470.2
XP_011541772.1
9 - 10
990
62
XM_011543471.1
XP_011541773.1
8 - 9
943
62
XM_011543473.1
XP_011541775.1
8 - 9
891
62
XM_011543477.2
XP_011541779.1
8 - 9
893
62
XM_017009565.1
XP_016865054.1
9 - 10
1103
62
XM_017009566.1
XP_016865055.1
9 - 10
993
62
XM_017009567.1
XP_016865056.1
7 - 8
731
62
XM_017009568.1
XP_016865057.1
9 - 10
977
62
XM_017009569.1
XP_016865058.1
8 - 9
856
62
XM_017009570.1
XP_016865059.1
5 - 6
1676
62
XM_017009571.1
XP_016865060.1
8 - 9
899
62
GenBank mRNA
AF012073.1
-
6 - 7
839
62
AF012074.1
-
2 - 3
575
62
AF536976.1
-
8 - 9
800
62
AF536977.1
-
6 - 7
603
62
AJ250852.1
-
2 - 3
171
62
AJ250854.1
-
7 - 8
766
62
AY245866.1
-
8 - 9
942
62
AY245867.1
-
6 - 7
1241
62
AY388960.1
-
6 - 7
524
62
L20970.1
-
6 - 7
637
62
U02882.1
-
5 - 6
407
62
U50157.1
-
1 - 2
245
62
U50158.1
-
2 - 3
159
62
U50159.1
-
6 - 7
580
62
U79571.1
-
1 - 2
650
62
Back To Top
More Information
Additional Information:
This assay is in the 5'UTR region of the AF012074.1, NM_001197221.1, U50158.1 transcripts.
Set Membership:
Ambion siRNA available
Amplicon length less than or equal to 70
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene PDE4D
Gene Ontology Categories:
Function(s)
Process(es)
regulation of heart rate
cAMP catabolic process
smooth muscle contraction
aging
regulation of signaling receptor activity
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
cAMP-mediated signaling
neutrophil chemotaxis
positive regulation of interferon-gamma production
positive regulation of interleukin-2 production
positive regulation of interleukin-5 production
negative regulation of peptidyl-serine phosphorylation
multicellular organism growth
negative regulation of heart contraction
T cell receptor signaling pathway
regulation of ryanodine-sensitive calcium-release channel activity
establishment of endothelial barrier
cellular response to lipopolysaccharide
cellular response to epinephrine stimulus
adrenergic receptor signaling pathway
regulation of cardiac muscle cell contraction
adenylate cyclase-activating adrenergic receptor signaling pathway involved in positive regulation of heart rate
regulation of cell communication by electrical coupling involved in cardiac conduction
negative regulation of relaxation of cardiac muscle
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
protein binding
enzyme binding
cAMP binding
beta-2 adrenergic receptor binding
transmembrane transporter binding
metal ion binding
ATPase binding
scaffold protein binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0