Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00939737_g1
          See other EDA2R GE Assays ›
          Gene Symbol:
          EDA2R
          Gene Name:
          ectodysplasin A2 receptor
          Gene Aliases:
          EDA-A2R, EDAA2R, TNFRSF27, XEDAR
          Chromosome Location:
          Chr.X: 66594384 - 66639303 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00939737_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: EDA2R
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001199687.2 NP_001186616.1 3 - 4 369 72
          NM_001242310.1 NP_001229239.1 2 - 3 271 72
          NM_001324206.1 NP_001311135.1 3 - 4 371 72
          NM_021783.4 NP_068555.1 3 - 4 371 72
          NR_136726.1 3 - 4 369 72
          NR_136727.1 3 - 4 371 72
          XM_011531000.1 XP_011529302.1 3 - 4 321 72
          XM_011531001.1 XP_011529303.1 3 - 4 369 72
          XM_011531002.1 XP_011529304.1 3 - 4 321 72
          XM_011531004.1 XP_011529306.1 3 - 4 321 72
          XM_011531006.2 XP_011529308.1 3 - 4 361 72
          XM_011531007.2 XP_011529309.1 3 - 4 361 72
          XM_017029704.1 XP_016885193.1 3 - 4 321 72
          XM_017029706.1 XP_016885195.1 3 - 4 321 72
          GenBank mRNA AF298812.1 - 2 - 3 261 72
          AK296264.1 - 3 - 4 321 72
          AK314881.1 - 3 - 4 337 72
          AY152724.1 - 2 - 3 261 72
          AY358735.1 - 2 - 3 264 72
          AY358736.1 - 2 - 3 264 72
          BC034919.1 - 3 - 4 318 5434545 72
          BP249106.1 - 3 - 4 337 72
          DA215848.1 - 3 - 4 370 72
          DA574050.1 - 3 - 4 321 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00009917.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X001101.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene EDA2R

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          epidermis development
          tissue development
          ectodermal cell differentiation
          tumor necrosis factor-mediated signaling pathway
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of JNK cascade
          positive regulation of NF-kappaB transcription factor activity
          intrinsic apoptotic signaling pathway by p53 class mediator
          tumor necrosis factor-activated receptor activity
          protein binding
          signaling receptor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X015022.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X015022.hg.1 ENST00000374719.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X015022.hg.1 ENST00000396050.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X015022.hg.1 ENST00000450752.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X015022.hg.1 ENST00000456230.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X015022.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X015022.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X015022.hg.1 NM_021783.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X015022.hg.1 TC0X001101.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Clariom D Assay PSR0X00167756.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00167756.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00167756.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00167756.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00167756.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00167756.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00167756.hg.1 NM_021783.4 Refseq
          Affymetrix Human Clariom D Assay PSR0X00167756.hg.1 TC0X00009917.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00167756.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00167756.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00167756.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00167756.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00167756.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00167756.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00167756.hg.1 NM_021783.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0