Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00939745_g1
          See other TRAF6 GE Assays ›
          Gene Symbol:
          TRAF6
          Gene Name:
          TNF receptor associated factor 6
          Gene Aliases:
          MGC:3310, RNF85
          Chromosome Location:
          Chr.11: 36483767 - 36510313 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00939745_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TRAF6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004620.3 NP_004611.1 4 - 5 897 74
          NM_145803.2 NP_665802.1 5 - 6 990 74
          GenBank mRNA AK292978.1 - 4 - 5 856 74
          BC031052.1 - 5 - 6 935 5272008 74
          U78798.1 - 4 - 5 830 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100010551.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11001560.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TRAF6

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          protein polyubiquitination
          ossification
          neural tube closure
          stimulatory C-type lectin receptor signaling pathway
          toll-like receptor signaling pathway
          regulation of immunoglobulin production
          positive regulation of T cell cytokine production
          MyD88-dependent toll-like receptor signaling pathway
          MyD88-independent toll-like receptor signaling pathway
          I-kappaB kinase/NF-kappaB signaling
          activation of NF-kappaB-inducing kinase activity
          JNK cascade
          antigen processing and presentation of exogenous peptide antigen via MHC class II
          osteoclast differentiation
          membrane protein intracellular domain proteolysis
          positive regulation of protein ubiquitination
          positive regulation of lipopolysaccharide-mediated signaling pathway
          activation of protein kinase activity
          positive regulation of interleukin-2 production
          toll-like receptor 9 signaling pathway
          Fc-epsilon receptor signaling pathway
          T-helper 1 type immune response
          positive regulation of T cell proliferation
          odontogenesis of dentin-containing tooth
          myeloid dendritic cell differentiation
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of JUN kinase activity
          bone resorption
          positive regulation of osteoclast differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          cell development
          positive regulation of smooth muscle cell proliferation
          T cell receptor signaling pathway
          positive regulation of T cell activation
          positive regulation of DNA-binding transcription factor activity
          positive regulation of NF-kappaB transcription factor activity
          protein autoubiquitination
          protein-containing complex assembly
          nucleotide-binding oligomerization domain containing signaling pathway
          interleukin-1-mediated signaling pathway
          protein K63-linked ubiquitination
          response to interleukin-1
          cellular response to lipopolysaccharide
          positive regulation of transcription regulatory region DNA binding
          ubiquitin-protein transferase activity
          tumor necrosis factor receptor binding
          protein binding
          zinc ion binding
          ligase activity
          protein kinase binding
          mitogen-activated protein kinase kinase kinase binding
          ubiquitin conjugating enzyme binding
          ubiquitin protein ligase binding
          thioesterase binding
          identical protein binding
          histone deacetylase binding
          protein kinase B binding
          protein N-terminus binding
          ubiquitin protein ligase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR11019578.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11019578.hg.1 ENST00000526995.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11019578.hg.1 ENST00000529150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11019578.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11019578.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11019578.hg.1 TC11001560.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000529150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000529150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000529150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000529150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000529150.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000529150.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000529150.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000529150.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100180082.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100180082.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100180082.hg.1 ENST00000529150.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100180082.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100180082.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100180082.hg.1 TC1100010551.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100180082.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100180082.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100180082.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100180082.hg.1 ENST00000529150.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100180082.hg.1 NM_145803.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0