Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00939897_g1
          See other F11R GE Assays ›
          Gene Symbol:
          F11R
          Gene Name:
          F11 receptor
          Gene Aliases:
          CD321, JAM, JAM1, JAMA, JCAM, KAT, PAM-1
          Chromosome Location:
          Chr.1: 160995211 - 161021343 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00939897_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: F11R
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016946.4 NP_058642.1 8 - 9 1085 63
          GenBank mRNA AF154005.1 - 8 - 9 757 63
          AF172398.2 - 8 - 9 961 63
          AF207907.1 - 8 - 9 830 63
          AK026665.1 - 8 - 9 876 63
          AK075152.1 - 8 - 9 885 63
          AK299487.1 - 8 - 9 866 63
          AK304412.1 - 7 - 8 691 63
          AL136649.1 - 8 - 9 889 63
          AY358896.1 - 8 - 9 866 63
          BC001533.2 - 8 - 9 859 2990019 63
          BT020103.1 - 8 - 9 815 63
          CR533512.1 - 8 - 9 815 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100018519.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01003412.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene F11R

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cytokine production
          inflammatory response
          transforming growth factor beta receptor signaling pathway
          response to radiation
          extracellular matrix organization
          actomyosin structure organization
          negative regulation of GTPase activity
          positive regulation of GTPase activity
          positive regulation of blood pressure
          viral entry into host cell
          intestinal absorption
          leukocyte migration
          bicellular tight junction assembly
          protein localization to plasma membrane
          establishment of endothelial intestinal barrier
          regulation of membrane permeability
          cell-cell adhesion
          regulation of actin cytoskeleton reorganization
          virus receptor activity
          protein binding
          PDZ domain binding
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01053354.hg.1 ENST00000289779.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01053354.hg.1 ENST00000335772.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01053354.hg.1 ENST00000368026.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01053354.hg.1 ENST00000436182.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01053354.hg.1 ENST00000537746.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01053354.hg.1 NM_016946.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01053354.hg.1 TC01003412.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100243905.hg.1 ENST00000368026.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243905.hg.1 ENST00000537746.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243905.hg.1 ENST00000621309.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243905.hg.1 NM_016946.5 Refseq
          Affymetrix Human Clariom D Assay PSR0100243905.hg.1 TC0100018519.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100243905.hg.1 ENST00000368026.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243905.hg.1 ENST00000537746.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243905.hg.1 ENST00000621309.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243905.hg.1 NM_016946.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc