Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs00942033_g1
See other SYNE1 GE Assays ›
Gene Symbol:
SYNE1
Gene Name:
spectrin repeat containing nuclear envelope protein 1
Gene Aliases:
8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
Chromosome Location:
Chr.6: 152121684 - 152637399 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs00942033_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: SYNE1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_033071.3
NP_149062.1
6 - 7
766
77
XM_006715407.1
XP_006715470.1
6 - 7
808
77
XM_006715408.2
XP_006715471.1
6 - 7
812
77
XM_006715410.2
XP_006715473.1
6 - 7
812
77
XM_006715411.1
XP_006715474.1
6 - 7
808
77
XM_006715412.2
XP_006715475.1
6 - 7
812
77
XM_006715413.2
XP_006715476.1
6 - 7
812
77
XM_006715415.2
XP_006715478.1
6 - 7
812
77
XM_006715416.2
XP_006715479.1
6 - 7
812
77
XM_006715417.2
XP_006715480.1
6 - 7
812
77
XM_006715420.2
XP_006715483.1
6 - 7
812
77
XM_006715421.2
XP_006715484.1
6 - 7
812
77
XM_006715422.1
XP_006715485.1
6 - 7
808
77
XM_006715423.2
XP_006715486.1
6 - 7
812
77
XM_006715424.2
XP_006715487.1
6 - 7
812
77
XM_006715425.2
XP_006715488.1
6 - 7
812
77
XM_011535641.2
XP_011533943.1
6 - 7
812
77
XM_011535642.2
XP_011533944.1
6 - 7
812
77
XM_011535643.1
XP_011533945.1
6 - 7
808
77
XM_017010608.1
XP_016866097.1
7 - 8
927
77
XM_017010609.1
XP_016866098.1
6 - 7
734
77
XM_017010611.1
XP_016866100.1
5 - 6
516
77
XM_017010612.1
XP_016866101.1
5 - 6
491
77
XM_017010613.1
XP_016866102.1
6 - 7
812
77
XM_017010614.1
XP_016866103.1
6 - 7
812
77
XM_017010615.1
XP_016866104.1
6 - 7
812
77
XM_017010616.1
XP_016866105.1
6 - 7
812
77
XM_017010617.1
XP_016866106.1
6 - 7
812
77
XM_017010618.1
XP_016866107.1
6 - 7
812
77
GenBank mRNA
AF535142.1
-
6 - 7
766
77
JQ740783.1
-
7 - 8
931
77
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length between 71 and 85
Detect Genomic DNA
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene SYNE1
Gene Ontology Categories:
Function(s)
Process(es)
nucleus organization
Golgi organization
nuclear migration
muscle cell differentiation
nuclear matrix anchoring at nuclear membrane
RNA binding
actin binding
protein binding
lamin binding
protein homodimerization activity
actin filament binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0