Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00944785_g1
          See other CTNNA1 GE Assays ›
          Gene Symbol:
          CTNNA1
          Gene Name:
          catenin alpha 1
          Gene Aliases:
          CAP102, MDPT2
          Chromosome Location:
          Chr.5: 138753386 - 138935034 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00944785_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CTNNA1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001290307.2 NP_001277236.1 10 - 11 1515 79
          NM_001290309.2 NP_001277238.1 9 - 10 1319 79
          NM_001290310.2 NP_001277239.1 10 - 11 1352 79
          NM_001290312.1 NP_001277241.1 4 - 5 835 79
          NM_001323982.1 NP_001310911.1 11 - 12 1985 79
          NM_001323983.1 NP_001310912.1 10 - 11 1667 79
          NM_001323984.1 NP_001310913.1 11 - 12 1533 79
          NM_001323985.1 NP_001310914.1 10 - 11 1515 79
          NM_001323987.1 NP_001310916.1 4 - 5 524 79
          NM_001323988.1 NP_001310917.1 7 - 8 772 79
          NM_001323989.1 NP_001310918.1 5 - 6 620 79
          NM_001323990.1 NP_001310919.1 6 - 7 984 79
          NM_001323991.1 NP_001310920.1 5 - 6 931 79
          NM_001323992.1 NP_001310921.1 7 - 8 764 79
          NM_001323993.1 NP_001310922.1 7 - 8 1078 79
          NM_001323994.1 NP_001310923.1 4 - 5 533 79
          NM_001323995.1 NP_001310924.1 7 - 8 748 79
          NM_001323996.1 NP_001310925.1 7 - 8 1083 79
          NM_001323997.1 NP_001310926.1 7 - 8 767 79
          NM_001323998.1 NP_001310927.1 7 - 8 1059 79
          NM_001323999.1 NP_001310928.1 7 - 8 1056 79
          NM_001324000.1 NP_001310929.1 5 - 6 928 79
          NM_001324001.1 NP_001310930.1 4 - 5 835 79
          NM_001324002.1 NP_001310931.1 4 - 5 835 79
          NM_001324003.1 NP_001310932.1 6 - 7 984 79
          NM_001324004.1 NP_001310933.1 4 - 5 524 79
          NM_001324005.1 NP_001310934.1 7 - 8 1059 79
          NM_001324012.1 NP_001310941.1 2 - 3 290 79
          NM_001324013.1 NP_001310942.1 5 - 6 864 79
          NM_001903.4 NP_001894.2 10 - 11 1515 79
          GenBank mRNA AK094151.1 - 7 - 8 724 79
          AK296708.1 - 10 - 11 1323 79
          AK304267.1 - 10 - 11 1410 79
          BC000385.2 - 10 - 11 1453 2821111 79
          BC031262.1 - 4 - 5 414 5274351 79
          D13866.1 - 10 - 11 1454 79
          D14705.1 - 10 - 11 1479 79
          HQ589335.1 - 4 - 5 669 79

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the NM_001324012.1, NM_001324013.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CTNNA1

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle development
          actin filament organization
          cell adhesion
          establishment or maintenance of cell polarity
          negative regulation of neuroblast proliferation
          aging
          male gonad development
          gap junction assembly
          axon regeneration
          adherens junction organization
          cellular protein localization
          odontogenesis of dentin-containing tooth
          apical junction assembly
          response to estrogen
          positive regulation of smoothened signaling pathway
          positive regulation of muscle cell differentiation
          protein heterooligomerization
          cellular response to indole-3-methanol
          epithelial cell-cell adhesion
          negative regulation of cell motility
          negative regulation of integrin-mediated signaling pathway
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          RNA binding
          structural molecule activity
          protein binding
          beta-catenin binding
          vinculin binding
          gamma-catenin binding
          cadherin binding
          actin filament binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0