Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs00944801_g1
See other CTNNA1 GE Assays ›
Gene Symbol:
CTNNA1
Gene Name:
catenin alpha 1
Gene Aliases:
CAP102, MDPT2
Chromosome Location:
Chr.5: 138753386 - 138935034 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs00944801_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: CTNNA1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001290307.2
NP_001277236.1
9 - 10
1421
97
NM_001290309.2
NP_001277238.1
8 - 9
1225
97
NM_001290310.2
NP_001277239.1
9 - 10
1258
97
NM_001290312.1
NP_001277241.1
3 - 4
741
97
NM_001323982.1
NP_001310911.1
10 - 11
1891
97
NM_001323983.1
NP_001310912.1
9 - 10
1573
97
NM_001323984.1
NP_001310913.1
10 - 11
1439
97
NM_001323985.1
NP_001310914.1
9 - 10
1421
97
NM_001323987.1
NP_001310916.1
3 - 4
430
97
NM_001323988.1
NP_001310917.1
6 - 7
678
97
NM_001323989.1
NP_001310918.1
4 - 5
526
97
NM_001323990.1
NP_001310919.1
5 - 6
890
97
NM_001323991.1
NP_001310920.1
4 - 5
837
97
NM_001323992.1
NP_001310921.1
6 - 7
670
97
NM_001323993.1
NP_001310922.1
6 - 7
984
97
NM_001323994.1
NP_001310923.1
3 - 4
439
97
NM_001323995.1
NP_001310924.1
6 - 7
654
97
NM_001323996.1
NP_001310925.1
6 - 7
989
97
NM_001323997.1
NP_001310926.1
6 - 7
673
97
NM_001323998.1
NP_001310927.1
6 - 7
965
97
NM_001323999.1
NP_001310928.1
6 - 7
962
97
NM_001324000.1
NP_001310929.1
4 - 5
834
97
NM_001324001.1
NP_001310930.1
3 - 4
741
97
NM_001324002.1
NP_001310931.1
3 - 4
741
97
NM_001324003.1
NP_001310932.1
5 - 6
890
97
NM_001324004.1
NP_001310933.1
3 - 4
430
97
NM_001324005.1
NP_001310934.1
6 - 7
965
97
NM_001324006.1
NP_001310935.1
6 - 7
654
97
NM_001324007.1
NP_001310936.1
5 - 6
890
97
NM_001324008.1
NP_001310937.1
3 - 4
430
97
NM_001324009.1
NP_001310938.1
3 - 4
741
97
NM_001903.4
NP_001894.2
9 - 10
1421
97
GenBank mRNA
AK094151.1
-
6 - 7
630
97
AK296708.1
-
9 - 10
1229
97
AK300439.1
-
8 - 9
1192
97
AK304267.1
-
9 - 10
1316
97
BC000385.2
-
9 - 10
1359
2821111
97
BC031262.1
-
3 - 4
320
5274351
97
D13866.1
-
9 - 10
1360
97
D14705.1
-
9 - 10
1385
97
HQ589335.1
-
3 - 4
575
97
L23805.1
-
8 - 9
1298
97
U03100.1
-
8 - 9
1298
97
Back To Top
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001324006.1, NM_001324007.1, NM_001324008.1, NM_001324009.1 transcripts.
Set Membership:
Ambion siRNA available
Amplicon length between 86 and 100
Mammalian Gene Collection
Detect Genomic DNA
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene CTNNA1
Gene Ontology Categories:
Function(s)
Process(es)
ovarian follicle development
actin filament organization
cell adhesion
establishment or maintenance of cell polarity
negative regulation of neuroblast proliferation
aging
male gonad development
gap junction assembly
axon regeneration
adherens junction organization
cellular protein localization
odontogenesis of dentin-containing tooth
apical junction assembly
response to estrogen
positive regulation of smoothened signaling pathway
positive regulation of muscle cell differentiation
protein heterooligomerization
cellular response to indole-3-methanol
epithelial cell-cell adhesion
negative regulation of cell motility
negative regulation of integrin-mediated signaling pathway
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
RNA binding
structural molecule activity
protein binding
beta-catenin binding
vinculin binding
gamma-catenin binding
cadherin binding
actin filament binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0