Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00946084_g1
          See other HSPA5 GE Assays ›
          Gene Symbol:
          HSPA5
          Gene Name:
          heat shock protein family A (Hsp70) member 5
          Gene Aliases:
          BIP, GRP78, HEL-S-89n, MIF2
          Chromosome Location:
          Chr.9: 125234848 - 125241387 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00946084_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HSPA5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005347.4 NP_005338.1 3 - 4 755 87
          GenBank mRNA AF188611.1 - 3 - 4 440 87
          AF216292.1 - 3 - 4 494 87
          AJ271729.1 - 3 - 4 507 87
          AK293604.1 - 3 - 4 711 87
          BC020235.1 - 3 - 4 698 5020098 87
          EU794617.1 - 3 - 4 507 87
          X87949.1 - 3 - 4 716 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900011523.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09001589.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HSPA5

          Gene Ontology Categories:

          Function(s) Process(es)

          ER overload response
          activation of signaling protein activity involved in unfolded protein response
          cerebellum structural organization
          cerebellar Purkinje cell layer development
          substantia nigra development
          positive regulation of cell migration
          ER-associated ubiquitin-dependent protein catabolic process
          negative regulation of transforming growth factor beta receptor signaling pathway
          endoplasmic reticulum unfolded protein response
          positive regulation of protein ubiquitination
          protein folding in endoplasmic reticulum
          maintenance of protein localization in endoplasmic reticulum
          IRE1-mediated unfolded protein response
          PERK-mediated unfolded protein response
          ATF6-mediated unfolded protein response
          cellular response to glucose starvation
          negative regulation of apoptotic process
          regulation of protein folding in endoplasmic reticulum
          cellular response to antibiotic
          cellular response to manganese ion
          cellular response to interleukin-4
          negative regulation of protein homodimerization activity
          cell-cell adhesion
          toxin transport
          regulation of ATF6-mediated unfolded protein response
          regulation of IRE1-mediated unfolded protein response
          regulation of PERK-mediated unfolded protein response
          positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
          glycoprotein binding
          calcium ion binding
          protein binding
          ATP binding
          ATPase activity
          enzyme binding
          protein domain specific binding
          ubiquitin protein ligase binding
          ribosome binding
          unfolded protein binding
          chaperone binding
          misfolded protein binding
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100007873.hg.1 ENST00000442006.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007873.hg.1 ENST00000442006.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007873.hg.1 ENST00000442006.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007873.hg.1 ENST00000442006.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007873.hg.1 ENST00000442006.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011523.hg.1 ENST00000324460.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011523.hg.1 ENST00000324460.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011523.hg.1 ENST00000324460.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011523.hg.1 ENST00000324460.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011523.hg.1 ENST00000324460.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011523.hg.1 ENST00000324460.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011523.hg.1 ENST00000324460.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011523.hg.1 ENST00000324460.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011523.hg.1 ENST00000324460.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011523.hg.1 ENST00000324460.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011523.hg.1 NM_005347.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011523.hg.1 NM_005347.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011523.hg.1 NM_005347.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011523.hg.1 NM_005347.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011523.hg.1 NM_005347.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011523.hg.1 NM_005347.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011523.hg.1 NM_005347.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011523.hg.1 NM_005347.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011523.hg.1 NM_005347.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011523.hg.1 NM_005347.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011523.hg.1 NM_005347.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007873.hg.1 ENST00000442006.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007873.hg.1 ENST00000442006.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007873.hg.1 ENST00000442006.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007873.hg.1 ENST00000442006.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007873.hg.1 ENST00000442006.2 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0