Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00947391_m1
          See other PKD1 GE Assays ›
          Gene Symbol:
          PKD1
          Gene Name:
          polycystin 1, transient receptor potential channel interacting
          Gene Aliases:
          PBP, Pc-1, TRPP1
          Chromosome Location:
          Chr.16: 2088708 - 2135898 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00947391_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PKD1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000296.3 NP_000287.3 34 - 35 10710 73
          NM_001009944.2 NP_001009944.2 34 - 35 10713 73
          XM_005255370.3 XP_005255427.1 24 - 25 8300 73
          XM_011522528.2 XP_011520830.1 35 - 36 10738 73
          XM_011522529.2 XP_011520831.1 35 - 36 10736 73
          XM_011522530.2 XP_011520832.1 35 - 36 10543 73
          XM_011522532.2 XP_011520834.1 34 - 35 10471 73
          XM_011522534.2 XP_011520836.1 35 - 36 10528 73
          XM_011522535.2 XP_011520837.1 26 - 27 8760 73
          XM_011522537.2 XP_011520839.1 24 - 25 7671 73
          XM_017023285.1 XP_016878774.1 25 - 26 8315 73
          GenBank mRNA AB209025.1 - 21 - 22 5067 73
          AB209675.1 - 20 - 21 6019 73
          L33243.1 - 34 - 35 10712 73
          U24497.1 - 34 - 35 10639 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600009088.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC16000773.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PKD1

          Gene Ontology Categories:

          Function(s) Process(es)

          cartilage condensation
          in utero embryonic development
          kidney development
          liver development
          embryonic placenta development
          protein export from nucleus
          nitrogen compound metabolic process
          homophilic cell adhesion via plasma membrane adhesion molecules
          cell-matrix adhesion
          calcium-independent cell-matrix adhesion
          positive regulation of cytosolic calcium ion concentration
          receptor signaling pathway via JAK-STAT
          heart development
          anatomical structure morphogenesis
          peptidyl-serine phosphorylation
          spinal cord development
          neural tube development
          establishment of cell polarity
          regulation of cell adhesion
          positive regulation of protein binding
          response to fluid shear stress
          lymph vessel morphogenesis
          cytoplasmic sequestering of transcription factor
          skin development
          positive regulation of transcription by RNA polymerase II
          digestive tract development
          branching morphogenesis of an epithelial tube
          genitalia development
          detection of mechanical stimulus
          cartilage development
          regulation of mitotic spindle organization
          lung epithelium development
          placenta blood vessel development
          regulation of proteasomal protein catabolic process
          calcium ion transmembrane transport
          mesonephric tubule development
          mesonephric duct development
          metanephric collecting duct development
          metanephric ascending thin limb development
          metanephric proximal tubule development
          metanephric distal tubule morphogenesis
          cell-cell adhesion
          calcium channel activity
          protein binding
          protein kinase binding
          protein domain specific binding
          carbohydrate binding
          transmembrane transporter binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR16011309.hg.1 ENST00000262304.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16011309.hg.1 ENST00000306101.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16011309.hg.1 ENST00000382481.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16011309.hg.1 ENST00000423118.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16011309.hg.1 ENST00000487932.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16011309.hg.1 NM_000296.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16011309.hg.1 NM_001009944.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16011309.hg.1 TC16000773.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1600169039.hg.1 ENST00000262304.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1600169039.hg.1 ENST00000423118.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600169039.hg.1 ENST00000487932.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600169039.hg.1 NM_000296.3 Refseq
          Affymetrix Human Clariom D Assay PSR1600169039.hg.1 NM_001009944.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600169039.hg.1 TC1600009088.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600169039.hg.1 ENST00000262304.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1600169039.hg.1 ENST00000423118.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600169039.hg.1 ENST00000487932.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600169039.hg.1 NM_000296.3 Refseq
          Affymetrix Human Clariom D Assay PSR1600169039.hg.1 NM_001009944.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-tbs5g:80/100.66.134.240:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0