Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00947523_m1
          See other ASPH GE Assays ›
          Gene Symbol:
          ASPH
          Gene Name:
          aspartate beta-hydroxylase
          Gene Aliases:
          AAH, BAH, CASQ2BP1, FDLAB, HAAH, JCTN, junctin
          Chromosome Location:
          Chr.8: 61500556 - 61714640 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00947523_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ASPH
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164751.1 NP_001158223.1 3 - 4 367 83
          NM_001164752.1 NP_001158224.1 3 - 4 367 83
          NM_001164753.1 NP_001158225.1 3 - 4 367 83
          NM_001164756.1 NP_001158228.1 3 - 4 562 83
          NM_020164.4 NP_064549.1 3 - 4 367 83
          NM_032468.4 NP_115857.1 3 - 4 367 83
          XM_005251235.2 XP_005251292.1 3 - 4 531 83
          XM_005251236.2 XP_005251293.1 3 - 4 531 83
          XM_005251238.2 XP_005251295.1 3 - 4 531 83
          XM_005251239.2 XP_005251296.1 3 - 4 531 83
          XM_005251240.1 XP_005251297.1 3 - 4 309 83
          XM_005251243.2 XP_005251300.1 3 - 4 531 83
          XM_005251244.1 XP_005251301.1 3 - 4 309 83
          XM_005251247.2 XP_005251304.1 3 - 4 531 83
          XM_017013419.1 XP_016868908.1 3 - 4 531 83
          XM_017013420.1 XP_016868909.1 3 - 4 531 83
          XM_017013421.1 XP_016868910.1 3 - 4 531 83
          XM_017013423.1 XP_016868912.1 3 - 4 531 83
          XM_017013425.1 XP_016868914.1 3 - 4 531 83
          XM_017013426.1 XP_016868915.1 3 - 4 309 83
          XM_017013427.1 XP_016868916.1 3 - 4 309 83
          XM_017013430.1 XP_016868919.1 3 - 4 531 83
          XM_017013432.1 XP_016868921.1 3 - 4 531 83
          XM_017013434.1 XP_016868923.1 3 - 4 531 83
          XM_017013435.1 XP_016868924.1 3 - 4 309 83
          XM_017013436.1 XP_016868925.1 3 - 4 531 83
          XM_017013438.1 XP_016868927.1 3 - 4 531 83
          XM_017013440.1 XP_016868929.1 3 - 4 309 83
          XM_017013442.1 XP_016868931.1 3 - 4 531 83
          GenBank mRNA AF184241.1 - 3 - 4 247 83
          AF224469.1 - 3 - 4 292 83
          AF306765.1 - 3 - 4 288 83
          AK295528.1 - 3 - 4 331 83
          BC066929.1 - 3 - 4 458 83
          BC142967.1 - 3 - 4 439 83
          BC144362.1 - 3 - 4 297 83
          BC144363.1 - 3 - 4 298 83

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ASPH

          Gene Ontology Categories:

          Function(s) Process(es)

          muscle contraction
          pattern specification process
          negative regulation of cell population proliferation
          regulation of protein stability
          ion transmembrane transport
          limb morphogenesis
          peptidyl-aspartic acid hydroxylation
          positive regulation of proteolysis
          roof of mouth development
          face morphogenesis
          activation of cysteine-type endopeptidase activity
          regulation of protein depolymerization
          regulation of cardiac conduction
          structural molecule activity
          calcium ion binding
          protein binding
          structural constituent of muscle
          electron transfer activity
          peptidyl-aspartic acid 3-dioxygenase activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0