Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00947877_m1
          See other TGFBR2 GE Assays ›
          Gene Symbol:
          TGFBR2
          Gene Name:
          transforming growth factor beta receptor 2
          Gene Aliases:
          AAT3, FAA3, LDS1B, LDS2, LDS2B, MFS2, RIIC, TAAD2, TGFR-2, TGFbeta-RII
          Chromosome Location:
          Chr.3: 30606490 - 30694142 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00947877_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TGFBR2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024847.2 NP_001020018.1 4 - 5 917 98
          NM_003242.5 NP_003233.4 3 - 4 842 98
          XM_011534043.2 XP_011532345.1 4 - 5 690 98
          XM_011534045.2 XP_011532347.1 3 - 4 470 98
          XM_017007106.1 XP_016862595.1 3 - 4 2056 98
          GenBank mRNA AK300383.1 - 4 - 5 818 98
          AK304404.1 - 4 - 5 587 98
          AK314102.1 - 3 - 4 758 98
          BC040499.1 - 3 - 4 843 98
          D28131.1 - 4 - 5 475 98
          KC792576.1 - 3 - 4 499 98
          KC792577.1 - 3 - 4 499 98
          KC792578.1 - 3 - 4 499 98
          KC792579.1 - 3 - 4 499 98
          KC792580.1 - 3 - 4 499 98
          KU178359.1 - 3 - 4 460 98
          KU178360.1 - 4 - 5 535 98
          M85079.1 - 3 - 4 795 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300006925.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03000149.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TGFBR2

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel development
          branching involved in blood vessel morphogenesis
          vasculogenesis
          in utero embryonic development
          heart looping
          positive regulation of mesenchymal cell proliferation
          lens development in camera-type eye
          positive regulation of tolerance induction to self antigen
          positive regulation of B cell tolerance induction
          positive regulation of T cell tolerance induction
          outflow tract morphogenesis
          growth plate cartilage chondrocyte growth
          protein phosphorylation
          receptor-mediated endocytosis
          apoptotic process
          transforming growth factor beta receptor signaling pathway
          common-partner SMAD protein phosphorylation
          Notch signaling pathway
          smoothened signaling pathway
          gastrulation
          brain development
          heart development
          embryo implantation
          aging
          response to nutrient
          positive regulation of cell population proliferation
          response to xenobiotic stimulus
          response to mechanical stimulus
          response to glucose
          regulation of gene expression
          positive regulation of epithelial cell migration
          positive regulation of epithelial to mesenchymal transition
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          negative regulation of transforming growth factor beta receptor signaling pathway
          animal organ regeneration
          activation of protein kinase activity
          embryonic hemopoiesis
          wound healing
          regulation of cell population proliferation
          myeloid dendritic cell differentiation
          positive regulation of skeletal muscle tissue regeneration
          response to estrogen
          positive regulation of angiogenesis
          response to steroid hormone
          digestive tract development
          positive regulation of smooth muscle cell proliferation
          embryonic cranial skeleton morphogenesis
          positive regulation of NK T cell differentiation
          roof of mouth development
          negative regulation of cardiac muscle cell proliferation
          pathway-restricted SMAD protein phosphorylation
          ventricular septum morphogenesis
          bronchus morphogenesis
          trachea formation
          mammary gland morphogenesis
          lung lobe morphogenesis
          response to cholesterol
          positive regulation of epithelial to mesenchymal transition involved in endocardial cushion formation
          lens fiber cell apoptotic process
          positive regulation of reactive oxygen species metabolic process
          transmembrane receptor protein serine/threonine kinase activity
          transforming growth factor beta-activated receptor activity
          transforming growth factor beta receptor activity, type II
          protein binding
          ATP binding
          glycosaminoglycan binding
          mitogen-activated protein kinase kinase kinase binding
          type I transforming growth factor beta receptor binding
          SMAD binding
          metal ion binding
          transforming growth factor beta binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR03002713.hg.1 ENST00000295754.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03002713.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03002713.hg.1 ENST00000383765.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03002713.hg.1 ENST00000439925.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03002713.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03002713.hg.1 NM_003242.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03002713.hg.1 TC03000149.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03002714.hg.1 ENST00000295754.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03002714.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03002714.hg.1 ENST00000383765.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03002714.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03002714.hg.1 NM_003242.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03002714.hg.1 TC03000149.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_003242.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_003242.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_003242.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_003242.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_003242.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_003242.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_003242.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_003242.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_003242.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_003242.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_003242.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006925.hg.1 NM_003242.5 Refseq
          Affymetrix Human Clariom D Assay PSR0300148241.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0300148241.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300148241.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300148241.hg.1 NM_003242.5 Refseq
          Affymetrix Human Clariom D Assay PSR0300148241.hg.1 TC0300006925.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300148241.hg.1 NM_003242.5 Refseq
          Affymetrix Human Clariom D Assay PSR0300148241.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0300148241.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300148241.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300148242.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0300148242.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300148242.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300148242.hg.1 NM_003242.5 Refseq
          Affymetrix Human Clariom D Assay PSR0300148242.hg.1 TC0300006925.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300148242.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0300148242.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300148242.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300148242.hg.1 NM_003242.5 Refseq
          Affymetrix Human Clariom D Assay PSR0300148238.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0300148238.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300148238.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300148238.hg.1 NM_003242.5 Refseq
          Affymetrix Human Clariom D Assay PSR0300148238.hg.1 TC0300006925.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300148238.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0300148238.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300148238.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300148238.hg.1 NM_003242.5 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 ENST00000295754.9 Ensembl
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 ENST00000359013.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_001024847.2 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_003242.5 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_003242.5 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_003242.5 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_003242.5 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_003242.5 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_003242.5 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_003242.5 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_003242.5 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_003242.5 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_003242.5 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_003242.5 Refseq
          Affymetrix Human Clariom D Assay TC0300006925.hg.1 NM_003242.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0