Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038150.hg.1 |
ENST00000172853.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038150.hg.1 |
ENST00000397342.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038150.hg.1 |
ENST00000397345.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038150.hg.1 |
ENST00000409198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038150.hg.1 |
ENST00000427231.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038150.hg.1 |
NM_001164507.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038150.hg.1 |
NM_001164508.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038150.hg.1 |
NM_004543.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038150.hg.1 |
TC02002428.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
ENST00000172853.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
ENST00000397345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
ENST00000409198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
ENST00000413693.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
ENST00000427231.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
ENST00000603639.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
ENST00000604864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
ENST00000618972.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
NM_001164507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
NM_001164508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
NM_001271208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
NM_004543.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
TC0200014617.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
ENST00000413693.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
ENST00000172853.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
ENST00000397345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
ENST00000409198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
ENST00000427231.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
ENST00000603639.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
ENST00000604864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
ENST00000618972.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
NM_001164507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
NM_001164508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
NM_001271208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206185.hg.1 |
NM_004543.4 |
Refseq |