Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs00952539_m1
See other PLAGL1 GE Assays ›
Gene Symbol:
PLAGL1
Gene Name:
PLAG1 like zinc finger 1
Gene Aliases:
LOT1, ZAC, ZAC1
Chromosome Location:
Chr.6: 143940300 - 144064599 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs00952539_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: PLAGL1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001080951.2
NP_001074420.1
7 - 8
915
90
NM_001080952.2
NP_001074421.1
6 - 7
1067
90
NM_001080953.2
NP_001074422.1
6 - 7
1021
90
NM_001080954.2
NP_001074423.1
6 - 7
1020
90
NM_001289042.1
NP_001275971.1
4 - 5
711
90
NM_001289043.1
NP_001275972.1
6 - 7
834
90
NM_001289044.1
NP_001275973.1
5 - 6
785
90
NM_001289045.1
NP_001275974.1
6 - 7
1329
90
NM_001289046.1
NP_001275975.1
8 - 9
1310
90
NM_001289047.1
NP_001275976.1
3 - 4
874
90
NM_001289048.1
NP_001275977.1
6 - 7
1035
90
NM_001289049.1
NP_001275978.1
5 - 6
980
90
NM_001317156.1
NP_001304085.1
3 - 4
1241
90
NM_001317157.1
NP_001304086.1
2 - 3
1417
90
NM_001317159.1
NP_001304088.1
7 - 8
901
90
NM_001317161.1
NP_001304090.1
6 - 7
1035
90
NM_001317162.1
NP_001304091.1
7 - 8
1061
90
NM_006718.4
NP_006709.2
6 - 7
1053
90
GenBank mRNA
AJ006354.1
-
2 - 3
1447
90
AJ311395.1
-
8 - 9
1300
90
AK091707.1
-
3 - 4
1241
90
AK096331.1
-
6 - 7
1297
90
AK315184.1
-
3 - 4
575
90
BC074814.2
-
1 - 2
200
30915533
90
BC109085.2
-
2 - 3
507
40034491
90
BC109086.2
-
2 - 3
507
40034497
90
U72621.4
-
5 - 6
2392
90
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length between 86 and 100
3' Most
Mammalian Gene Collection
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene PLAGL1
Gene Ontology Categories:
Function(s)
Process(es)
transcription by RNA polymerase II
apoptotic process
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
cell differentiation
skeletal muscle cell differentiation
positive regulation of transcription by RNA polymerase II
RNA polymerase II transcription regulatory region sequence-specific DNA binding
DNA-binding transcription activator activity, RNA polymerase II-specific
DNA binding
metal ion binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0