Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00954037_g1
          See other ID3 GE Assays ›
          Gene Symbol:
          ID3
          Gene Name:
          inhibitor of DNA binding 3, HLH protein
          Gene Aliases:
          HEIR-1, bHLHb25
          Chromosome Location:
          Chr.1: 23557930 - 23559794 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00954037_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ID3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002167.4 NP_002158.3 1 - 2 135
          GenBank mRNA A17546.1 - 1 - 2 424 135
          AK290003.1 - 1 - 2 406 135
          BC003107.1 - 1 - 2 387 3543936 135
          BM921819.1 - 700 135
          X66924.1 - 1 - 2 424 135
          X69111.1 - 1 - 2 668 135

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100013293.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002347.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ID3

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          metanephros development
          regulation of DNA replication
          transcription, DNA-templated
          multicellular organism development
          central nervous system development
          heart development
          muscle organ development
          circadian rhythm
          response to wounding
          neuron differentiation
          epithelial cell differentiation
          notochord development
          odontogenesis
          positive regulation of apoptotic process
          negative regulation of DNA-binding transcription factor activity
          negative regulation of myoblast differentiation
          negative regulation of osteoblast differentiation
          negative regulation of transcription, DNA-templated
          regulation of cell cycle
          cellular response to leptomycin B
          DNA-binding transcription factor activity
          transcription corepressor activity
          protein binding
          transcription factor binding
          protein domain specific binding
          protein dimerization activity
          leptomycin B binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01036183.hg.1 ENST00000374561.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01036183.hg.1 ENST00000486541.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01036183.hg.1 NM_002167.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01036183.hg.1 TC01002347.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01036185.hg.1 ENST00000374561.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01036185.hg.1 NM_002167.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01036185.hg.1 TC01002347.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01036181.hg.1 ENST00000374561.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01036181.hg.1 ENST00000486541.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01036181.hg.1 NM_002167.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01036181.hg.1 TC01002347.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01036184.hg.1 ENST00000374561.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01036184.hg.1 NM_002167.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01036184.hg.1 TC01002347.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100013293.hg.1 ENST00000374561.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013293.hg.1 ENST00000374561.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013293.hg.1 ENST00000374561.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013293.hg.1 ENST00000374561.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013293.hg.1 ENST00000374561.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013293.hg.1 ENST00000374561.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013293.hg.1 ENST00000463312.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013293.hg.1 ENST00000463312.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013293.hg.1 ENST00000463312.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013293.hg.1 ENST00000463312.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013293.hg.1 ENST00000486541.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013293.hg.1 ENST00000486541.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013293.hg.1 ENST00000486541.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013293.hg.1 ENST00000486541.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013293.hg.1 NM_002167.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013293.hg.1 NM_002167.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013293.hg.1 NM_002167.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013293.hg.1 NM_002167.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013293.hg.1 NM_002167.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013293.hg.1 NM_002167.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100198581.hg.1 ENST00000374561.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198581.hg.1 ENST00000486541.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198581.hg.1 NM_002167.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100198581.hg.1 TC0100013293.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100198581.hg.1 NM_002167.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100198581.hg.1 ENST00000374561.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198581.hg.1 ENST00000486541.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198584.hg.1 ENST00000374561.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198584.hg.1 NM_002167.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100198584.hg.1 TC0100013293.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100198584.hg.1 ENST00000374561.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198584.hg.1 NM_002167.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100198586.hg.1 ENST00000374561.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198586.hg.1 NM_002167.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100198586.hg.1 TC0100013293.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100198586.hg.1 ENST00000374561.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198586.hg.1 NM_002167.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100198579.hg.1 ENST00000374561.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198579.hg.1 ENST00000463312.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198579.hg.1 ENST00000486541.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198579.hg.1 NM_002167.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100198579.hg.1 TC0100013293.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100198579.hg.1 ENST00000374561.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198579.hg.1 ENST00000463312.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198579.hg.1 ENST00000486541.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198579.hg.1 NM_002167.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013293.hg.1 ENST00000374561.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013293.hg.1 ENST00000374561.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013293.hg.1 ENST00000374561.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013293.hg.1 ENST00000374561.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013293.hg.1 ENST00000374561.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013293.hg.1 ENST00000374561.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013293.hg.1 ENST00000463312.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013293.hg.1 ENST00000463312.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013293.hg.1 ENST00000463312.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013293.hg.1 ENST00000463312.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013293.hg.1 ENST00000486541.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013293.hg.1 ENST00000486541.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013293.hg.1 ENST00000486541.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013293.hg.1 ENST00000486541.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013293.hg.1 NM_002167.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013293.hg.1 NM_002167.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013293.hg.1 NM_002167.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013293.hg.1 NM_002167.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013293.hg.1 NM_002167.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013293.hg.1 NM_002167.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 196116 - -
          ABI Human Genome Survey Microarray Version 2.0 235147 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix