Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00958054_gH
          See other ATP1A3 GE Assays ›
          Gene Symbol:
          ATP1A3
          Gene Name:
          ATPase Na+/K+ transporting subunit alpha 3
          Gene Aliases:
          AHC2, ATP1A1, CAPOS, DYT12, RDP
          Chromosome Location:
          Chr.19: 41966582 - 41994276 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00958054_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ATP1A3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001256213.1 NP_001243142.1 7 - 8 790 99
          NM_001256214.1 NP_001243143.1 7 - 8 960 99
          NM_152296.4 NP_689509.1 7 - 8 921 99
          GenBank mRNA AK122693.1 - 1 - 2 43 99
          AK223569.1 - 7 - 8 872 99
          AK293784.1 - 6 - 7 572 99
          AK295078.1 - 7 - 8 914 99
          AK295833.1 - 7 - 8 775 99
          AK296557.1 - 7 - 8 790 99
          AK307921.1 - 7 - 8 868 99
          AK316069.1 - 7 - 8 907 99
          BC009282.2 - 7 - 8 837 4125669 99
          BC009394.2 - 7 - 8 837 4135506 99
          BC015566.2 - 7 - 8 837 4097948 99
          BX478337.1 - 2 - 3 136 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900010782.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19001563.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ATP1A3

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular sodium ion homeostasis
          memory
          adult locomotory behavior
          visual learning
          response to xenobiotic stimulus
          establishment or maintenance of transmembrane electrochemical gradient
          cellular potassium ion homeostasis
          ion transmembrane transport
          ionotropic glutamate receptor signaling pathway
          sodium ion export across plasma membrane
          cardiac muscle contraction
          cellular response to steroid hormone stimulus
          regulation of cardiac muscle cell membrane potential
          cell communication by electrical coupling involved in cardiac conduction
          response to glycoside
          regulation of cardiac conduction
          potassium ion import across plasma membrane
          P-type sodium:potassium-exchanging transporter activity
          ATP binding
          D1 dopamine receptor binding
          heparan sulfate proteoglycan binding
          metal ion binding
          chaperone binding
          P-type sodium:potassium-exchanging transporter activity involved in regulation of cardiac muscle cell membrane potential
          steroid hormone binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR19021281.hg.1 TC19001563.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19021281.hg.1 ENST00000302102.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19021281.hg.1 ENST00000368080.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19021281.hg.1 ENST00000535899.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19021281.hg.1 ENST00000543770.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19021281.hg.1 ENST00000545399.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19021281.hg.1 NM_001256213.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19021281.hg.1 NM_001256214.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19021281.hg.1 NM_152296.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19021280.hg.1 TC19001563.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19021280.hg.1 ENST00000302102.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19021280.hg.1 ENST00000368080.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19021280.hg.1 ENST00000535899.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19021280.hg.1 ENST00000543770.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19021280.hg.1 ENST00000545399.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19021280.hg.1 NM_001256213.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19021280.hg.1 NM_001256214.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19021280.hg.1 NM_152296.4 Refseq
          Affymetrix Human Clariom D Assay PSR1900189111.hg.1 ENST00000302102.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189111.hg.1 ENST00000441343.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189111.hg.1 ENST00000485672.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189111.hg.1 ENST00000543770.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189111.hg.1 ENST00000545399.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189111.hg.1 ENST00000602133.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189111.hg.1 NM_001256213.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900189111.hg.1 NM_001256214.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900189111.hg.1 NM_152296.4 Refseq
          Affymetrix Human Clariom D Assay PSR1900189111.hg.1 TC1900010782.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900189111.hg.1 ENST00000302102.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189111.hg.1 ENST00000441343.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189111.hg.1 ENST00000485672.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189111.hg.1 ENST00000543770.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189111.hg.1 ENST00000545399.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189111.hg.1 ENST00000602133.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189111.hg.1 NM_001256213.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900189111.hg.1 NM_001256214.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900189111.hg.1 NM_152296.4 Refseq
          Affymetrix Human Clariom D Assay PSR1900189112.hg.1 ENST00000302102.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189112.hg.1 ENST00000441343.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189112.hg.1 ENST00000473086.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189112.hg.1 ENST00000485672.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189112.hg.1 ENST00000543770.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189112.hg.1 ENST00000545399.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189112.hg.1 ENST00000602133.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189112.hg.1 NM_001256213.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900189112.hg.1 NM_001256214.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900189112.hg.1 NM_152296.4 Refseq
          Affymetrix Human Clariom D Assay PSR1900189112.hg.1 TC1900010782.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900189112.hg.1 ENST00000302102.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189112.hg.1 ENST00000441343.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189112.hg.1 ENST00000473086.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189112.hg.1 ENST00000485672.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189112.hg.1 ENST00000543770.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189112.hg.1 ENST00000545399.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189112.hg.1 ENST00000602133.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189112.hg.1 NM_001256213.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900189112.hg.1 NM_001256214.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900189112.hg.1 NM_152296.4 Refseq
          Affymetrix Human Clariom D Assay PSR1900189113.hg.1 ENST00000302102.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189113.hg.1 ENST00000441343.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189113.hg.1 ENST00000473086.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189113.hg.1 ENST00000485672.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189113.hg.1 ENST00000543770.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189113.hg.1 ENST00000545399.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189113.hg.1 ENST00000602133.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189113.hg.1 NM_001256213.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900189113.hg.1 NM_001256214.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900189113.hg.1 NM_152296.4 Refseq
          Affymetrix Human Clariom D Assay PSR1900189113.hg.1 TC1900010782.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900189113.hg.1 ENST00000302102.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189113.hg.1 ENST00000441343.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189113.hg.1 ENST00000473086.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189113.hg.1 ENST00000485672.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189113.hg.1 ENST00000543770.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189113.hg.1 ENST00000545399.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189113.hg.1 ENST00000602133.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900189113.hg.1 NM_001256213.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900189113.hg.1 NM_001256214.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900189113.hg.1 NM_152296.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0