Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00958620_m1
          See other HAVCR2 GE Assays ›
          Gene Symbol:
          HAVCR2
          Gene Name:
          hepatitis A virus cellular receptor 2
          Gene Aliases:
          CD366, HAVcr-2, KIM-3, TIM3, TIMD-3, TIMD3, Tim-3
          Chromosome Location:
          Chr.5: 157085832 - 157109237 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00958620_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HAVCR2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_032782.4 NP_116171.3
          GenBank mRNA AB924452.1 -
          AF251707.1 -
          AF450242.1 -
          AF450243.1 -
          AK027334.1 -
          AK314406.1 -
          AY069944.1 -
          BC063431.1 - 5229539
          JX049979.1 -

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500013388.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05003434.hg.1

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          macrophage activation involved in immune response
          natural killer cell tolerance induction
          regulation of tolerance induction dependent upon immune response
          negative regulation of T-helper 1 type immune response
          negative regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
          inflammatory response
          negative regulation of gene expression
          negative regulation of myeloid dendritic cell activation
          negative regulation of NF-kappaB transcription factor activity
          negative regulation of interferon-alpha production
          negative regulation of interferon-gamma production
          negative regulation of interleukin-2 production
          negative regulation of interleukin-3 production
          negative regulation of interleukin-6 production
          negative regulation of tumor necrosis factor production
          positive regulation of chemokine production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-1 production
          positive regulation of interleukin-4 production
          positive regulation of tumor necrosis factor production
          negative regulation of natural killer cell activation
          toll-like receptor 3 signaling pathway
          toll-like receptor 7 signaling pathway
          toll-like receptor 9 signaling pathway
          positive regulation of T cell proliferation
          negative regulation of T cell proliferation
          positive regulation of macrophage activation
          innate immune response
          defense response to Gram-positive bacterium
          maternal process involved in female pregnancy
          positive regulation of ERK1 and ERK2 cascade
          cellular response to lipopolysaccharide
          negative regulation of granulocyte colony-stimulating factor production
          negative regulation of defense response to bacterium
          positive regulation of defense response to bacterium
          positive regulation of NIK/NF-kappaB signaling
          negative regulation of immunological synapse formation
          negative regulation of T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
          protein binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000517358.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000517358.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000521665.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000521665.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000521665.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000521665.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000521665.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000521665.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000522593.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000522593.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000522593.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000522593.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000522593.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000522593.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000522593.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000522593.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000522593.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000522902.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000522902.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000522902.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000522902.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 ENST00000522902.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 NM_032782.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 NM_032782.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 NM_032782.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 NM_032782.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 NM_032782.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 NM_032782.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 NM_032782.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 NM_032782.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 NM_032782.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 NM_032782.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 NM_032782.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013388.hg.1 NM_032782.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05027556.hg.1 ENST00000307851.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05027556.hg.1 ENST00000522593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05027556.hg.1 NM_032782.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05027556.hg.1 TC05003434.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0500197277.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0500197277.hg.1 ENST00000521665.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500197277.hg.1 ENST00000522593.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500197277.hg.1 NM_032782.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500197277.hg.1 TC0500013388.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500197277.hg.1 NM_032782.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500197277.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0500197277.hg.1 ENST00000521665.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500197277.hg.1 ENST00000522593.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000307851.8 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000517358.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000517358.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000521665.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000521665.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000521665.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000521665.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000521665.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000521665.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000522593.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000522593.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000522593.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000522593.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000522593.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000522593.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000522593.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000522593.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000522593.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000522902.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000522902.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000522902.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000522902.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 ENST00000522902.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 NM_032782.4 Refseq
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 NM_032782.4 Refseq
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 NM_032782.4 Refseq
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 NM_032782.4 Refseq
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 NM_032782.4 Refseq
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 NM_032782.4 Refseq
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 NM_032782.4 Refseq
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 NM_032782.4 Refseq
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 NM_032782.4 Refseq
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 NM_032782.4 Refseq
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 NM_032782.4 Refseq
          Affymetrix Human Clariom D Assay TC0500013388.hg.1 NM_032782.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0