Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003403.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003403.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003403.hg.1 |
TC09000323.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
ENST00000460688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
ENST00000460688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
ENST00000460688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
ENST00000460688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
ENST00000460688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
ENST00000460688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
ENST00000460688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
ENST00000460688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007518.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900150944.hg.1 |
ENST00000377182.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900150944.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900150944.hg.1 |
TC0900007518.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900150944.hg.1 |
NM_153267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900150944.hg.1 |
ENST00000377182.4 |
Ensembl |