Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00962420_g1
          See other TUBB GE Assays ›
          Gene Symbol:
          TUBB
          Gene Name:
          tubulin beta class I
          Gene Aliases:
          CDCBM6, CSCSC1, M40, OK/SW-cl.56, TUBB1, TUBB5
          Chromosome Location:
          Chr.6: 30720201 - 30725426 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00962420_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TUBB
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001293212.1 NP_001280141.1 2 - 3 562 62
          NM_001293213.1 NP_001280142.1 2 - 3 478 62
          NM_001293215.1 NP_001280144.1 2 - 3 465 62
          NM_001293216.1 NP_001280145.1 2 - 3 364 62
          NM_178014.3 NP_821133.1 2 - 3 478 62
          NR_120608.1 2 - 3 478 62
          GenBank mRNA AB062393.1 - 2 - 3 304 62
          AF070561.1 - 2 - 3 299 62
          AF070593.1 - 2 - 3 299 62
          AF070600.1 - 2 - 3 316 62
          AF141349.1 - 2 - 3 316 62
          AK098772.1 - 2 - 3 323 62
          AK297489.1 - 2 - 3 232 62
          AK297638.1 - 2 - 3 327 62
          AK298889.1 - 2 - 3 368 62
          AK302314.1 - 2 - 3 431 62
          AK316265.1 - 2 - 3 235 62
          AK316316.1 - 2 - 3 360 62
          BC001938.1 - 2 - 3 303 2820585 62
          BC002347.2 - 2 - 3 308 2964461 62
          BC005838.2 - 2 - 3 303 2820585 62
          BC007605.1 - 2 - 3 308 3350604 62
          BC013374.2 - 2 - 3 299 3946253 62
          BC019924.1 - 2 - 3 296 4998571 62
          BC020946.1 - 2 - 3 337 4795692 62
          BC021909.1 - 2 - 3 298 3509417 62
          BC062532.1 - 2 - 3 296 62
          BC070326.1 - 2 - 3 314 6698679 62
          BC103746.1 - 2 - 3 321 6503977 62
          BE245819.1 - 2 - 3 332 62
          BU557299.1 - 2 - 3 478 62
          DC405540.1 - 2 - 3 275 62
          V00599.1 - 1 - 2 151 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600007544.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000347.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK298889.1, AK316316.1, BC062532.1, NM_001293215.1, NM_001293216.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TUBB

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          movement of cell or subcellular component
          microtubule-based process
          cellular process
          cytoskeleton-dependent intracellular transport
          natural killer cell mediated cytotoxicity
          spindle assembly
          cell division
          GTPase activity
          structural molecule activity
          structural constituent of cytoskeleton
          protein binding
          GTP binding
          protein domain specific binding
          ubiquitin protein ligase binding
          GTPase activating protein binding
          MHC class I protein binding
          protein-containing complex binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR06003072.hg.1 ENST00000327892.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06003072.hg.1 ENST00000330914.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06003072.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06003072.hg.1 ENST00000396389.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06003072.hg.1 NM_178014.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06003072.hg.1 TC06000347.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 NM_001293214.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 NM_001293214.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 NM_001293214.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 NM_001293214.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 NM_001293214.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 NM_001293214.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 NM_001293214.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 NM_001293214.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 NM_001293214.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 NM_001293214.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007544.hg.1 NM_001293214.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600151187.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Clariom D Assay PSR0600151187.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600151187.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600151187.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600151187.hg.1 NM_001293214.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600151187.hg.1 TC0600007544.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600151187.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600151187.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Clariom D Assay PSR0600151187.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600151187.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600151187.hg.1 NM_001293214.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000327892.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000330914.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396384.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 ENST00000396389.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 NM_001293214.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 NM_001293214.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 NM_001293214.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 NM_001293214.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 NM_001293214.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 NM_001293214.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 NM_001293214.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 NM_001293214.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 NM_001293214.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 NM_001293214.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007544.hg.1 NM_001293214.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline