Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00964855_m1
          See other TBCD GE Assays ›
          Gene Symbol:
          TBCD
          Gene Name:
          tubulin folding cofactor D
          Gene Aliases:
          SSD-1, tfcD
          Chromosome Location:
          Chr.17: 82752048 - 82943186 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00964855_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TBCD
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005993.4 NP_005984.3 22 - 23 2139 72
          XM_005256396.4 XP_005256453.1 21 - 22 2104 72
          XM_005256399.4 XP_005256456.1 10 - 11 936 72
          XM_005256404.4 XP_005256461.1 10 - 11 978 72
          XM_006722290.2 XP_006722353.1 21 - 22 2036 72
          XM_006722291.3 XP_006722354.1 10 - 11 1260 72
          XM_006722292.2 XP_006722355.1 8 - 9 672 72
          XM_011523589.1 XP_011521891.1 17 - 18 1679 72
          XM_011523591.1 XP_011521893.1 17 - 18 1664 72
          XM_011523593.1 XP_011521895.1 16 - 17 1271 72
          XM_011523594.1 XP_011521896.1 9 - 10 741 72
          XM_011523595.2 XP_011521897.1 9 - 10 819 72
          XM_011523597.2 XP_011521899.1 7 - 8 1472 72
          XM_011523598.1 XP_011521900.1 7 - 8 530 72
          XM_011523599.2 XP_011521901.1 7 - 8 1248 72
          XM_011523600.2 XP_011521902.1 8 - 9 692 72
          XM_017024987.1 XP_016880476.1 22 - 23 2000 72
          XM_017024989.1 XP_016880478.1 6 - 7 386 72
          XM_017024990.1 XP_016880479.1 10 - 11 758 72
          GenBank mRNA AB023205.1 - 22 - 23 2118 72
          AK057975.1 - 7 - 8 646 72
          AK091959.1 - 21 - 22 2025 72
          AK293471.1 - 22 - 23 2117 72
          AL133562.1 - 18 - 19 1584 72
          BC003094.1 - 22 - 23 2139 3503192 72
          BC006364.1 - 22 - 23 2096 4131114 72
          BC039654.1 - 8 - 9 492 4865367 72
          BC150142.1 - 6 - 7 458 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as HTA transcript cluster(s) TC17000958.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TBCD

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          tubulin complex assembly
          post-chaperonin tubulin folding pathway
          negative regulation of cell-substrate adhesion
          negative regulation of microtubule polymerization
          adherens junction assembly
          positive regulation of GTPase activity
          bicellular tight junction assembly
          GTPase activator activity
          protein binding
          beta-tubulin binding
          chaperone binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR17012663.hg.1 ENST00000269368.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17012663.hg.1 ENST00000334614.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17012663.hg.1 ENST00000355528.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17012663.hg.1 ENST00000536182.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17012663.hg.1 ENST00000539345.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17012663.hg.1 NM_005993.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17012663.hg.1 TC17000958.hg.1 HTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0